Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fast Blue BB Salt hemi(zinc chloride) salt
Synonym(s):
4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, 4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt
Linear Formula:
C
17
H
18
N
3
O
3
Cl · 1/2 ZnCl
2
CAS Number:
5486-84-0
Molecular Weight:
415.94
EC Number:
226-817-4
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Chemical Composition
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F3378
Fast Blue BB Salt hemi(zinc chloride) salt,
Dye content ≥80 %, Powder
Expand
View Pricing
44670
Fast Blue BB Salt hemi(zinc chloride) salt,
suitable for microscopy
Expand
View Pricing