![Propionyl-L-carnitine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/244/155/1024dc71-7383-4b01-a016-8df1ea0a48bd/640/1024dc71-7383-4b01-a016-8df1ea0a48bd.png)
Propionyl-L-carnitine
![Propionyl-L-carnitine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/244/155/1024dc71-7383-4b01-a016-8df1ea0a48bd/640/1024dc71-7383-4b01-a016-8df1ea0a48bd.png)
Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium inner salt, L-Carnitine propionyl ester, C3-Carnitine, Propanoyl-L-carnitine
Empirical Formula (Hill Notation):
C10H19NO4
CAS Number:
Molecular Weight:
217.26
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing