Skip to Content
Merck
All Photos(1)

Documents

403A-7

Sigma-Aldrich

C3d Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (403A-74)
vial of 0.5 mL concentrate (403A-75)
bottle of 1.0 mL predilute (403A-77)
vial of 1.0 mL concentrate (403A-76)
bottle of 7.0 mL predilute (403A-78)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... CR2(1380)

General description

Complement component C3 is an integral part of the classical, lectin, and alternative complement pathways, and its cleavage products are involved in immune responses such as opsonization and cell lysis. C3d is a fragment of C3 that normally binds to pathogen surfaces to promote B-cell activation. However, deposition of C3d has also been observed in peritubular capillaries of transplanted kidneys undergoing acute allograft rejection.

Quality


IVD

IVD

IVD

RUO

Linkage

C3d Positive Control Slides, Product No. 403S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Bickerstaff et al.
The American journal of pathology, 173(2), 347-357 (2008-06-28)
Acute humoral rejection (AHR), which occurs in up to 8% of kidney transplant recipients, is a significant cause of renal allograft dysfunction and loss. More efficacious treatment modalities are needed to eliminate or curtail alloantibody production and its deleterious effects
Dirk R J Kuypers et al.
Transplantation, 76(1), 102-108 (2003-07-17)
C4d deposition in peritubular capillaries (PTCs) is a sign of humoral renal allograft rejection and an independent predictor of graft survival. Few investigators have focused on the meaning of capillary C3 deposition in rejecting grafts. Because C3 production can result
G Eggertsen et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 109(12), 825-834 (2002-02-16)
Among patients with early severe impairment of renal allograft function we have previously identified a group displaying isolated deposition of complement factor C3 in glomeruli. Here we studied the pattern of complement deposition more extensively in allograft biopsies from five
Katrin Pfaltz et al.
Journal of cutaneous pathology, 37(6), 654-658 (2009-10-30)
Direct immunofluorescence (DIF) testing is an important procedure in the diagnosis of autoimmune bullous dermatoses. We investigated the expression of C3d in formalin-fixed, paraffin-embedded tissue of autoimmune bullous dermatoses. The immunohistochemical expression of C3d in bullous pemphigoid (BP) (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service