Skip to Content
Merck
All Photos(2)

Documents

D5652

Sigma-Aldrich

Dulbecco′s Phosphate Buffered Saline

Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
eCl@ss:
32129211
NACRES:
NA.25

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

pH

7.2-7.8

solubility

water: 9.6 g/L, clear

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Phosphate Buffered Saline is a balanced salt solution (BSS) widely used in tissue culture.

Application

D-PBS is used to wash cells during preparation and serial transfer.
Dulbecco′s Phosphate Buffered Saline has been used:
  • as one of the pseudo-physiological solutions to study the in vitro degradation behavior of Fe–20Mn–1.2C alloy
  • to dissolve Angelica gigas powder for microencapsulation
  • to wash cells as part of cell culture treatments, wound-healing assay, transwell assay and apoptosis assay
  • to wash amniotic membrane for cellular analyses

Biochem/physiol Actions

Dulbecco′s Phosphate Buffered Saline serves as an irrigating, transporting, and diluting fluid while maintaining intra- and extracellular osmotic balance. It hydrates cells and acts as a source of bulk inorganic ions essential for normal cell metabolism. This buffered saline acts as the principal energy source for cell metabolism when combined with a carbohydrate, such as glucose. It provides a buffering system to maintain the medium within the physiological pH range.

Quantity

Formulated to contain 9.6 grams of powder per liter of medium.

Other Notes

This D-PBS is formulated without Ca or Mg. Formulations with Ca and Mg added are also available

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoxia Zhang et al.
Molecular vision, 24, 434-442 (2018-07-24)
Retinal G protein-coupled receptor (RGR) mRNA is transcribed in the outer nuclear layer of human retinas; however, it is not known whether the RGR gene is expressed in the rod or cone photoreceptors. In this study, we investigate broader expression
Ana C Bohórquez et al.
Journal of colloid and interface science, 506, 393-402 (2017-07-28)
The rotational diffusion of polyethylene glycol coated magnetic nanoparticles in serum albumin solutions was investigated in a range spanning 0mgmL
Functional enhancement of ultrafine Angelica gigas powder by spray-drying microencapsulation
Choi KO, et al.
LWT--Food Science and Technology, 101, 161-166 (2019)
Petr Heneberg et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 49(1), 54-67 (2018-10-06)
CD16 was previously suggested to be a new marker of basophils that is subject to downregulation by FcεRI crosslinking. Certain compounds, including supraoptimal concentrations of the PKC inhibitors, bisindolylmaleimides, decouple the release of granules containing CD203c, CD63 and histamine, and
Iqbal Hyder et al.
BMC molecular and cell biology, 21(1), 9-9 (2020-03-01)
Gene transfer by electroporation is an established method for the non-viral mediated transfection of mammalian cells. Primary cells pose a particular challenge for electroporation-mediated gene transfer, since they are more vulnerable than immortalized cells, and have a limited proliferative capacity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service