Skip to Content
Merck
All Photos(1)

Documents

31387

Sigma-Aldrich

Dextran from Leuconostoc spp.

Mr 15,000-25,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Leuconostoc spp)

form

crystalline
solid

mol wt

Mr 15,000-25,000

color

colorless

solubility

water: 0.5 g/5mL, colorless

storage temp.

room temp

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antitumor activity of sorafenib-incorporated nanoparticles of dextran/poly(DL-lactide-co-glycolide) block copolymer.
Kim DH, Kim MD, Choi CW, et al.
Nano Letters, 7, 91-91 (2012)
Yuichi Ozaki et al.
Circulation journal : official journal of the Japanese Circulation Society, 76(4), 922-927 (2012-02-04)
Although an intracoronary frequency-domain optical coherence tomography (FD-OCT) system overcomes several limitations of the time-domain OCT (TD-OCT) system, the former requires injection of contrast media for image acquisition. The increased total amount of contrast media for FD-OCT image acquisition may
Molecular Weight Effects on the Miscibility Behavior of Dextran and Maltodextrin with Poly(vinylpyrrolidone).
Van Eerdenbrugh B, Taylor LS.
Pharmaceut. Res., doi: 10-doi: 10 (2012)
Melda Altikatoglu et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 40(4), 261-265 (2012-01-28)
In the present study, the stabilizing effect of dextrans as additives on the denaturation and inactivation of glucose oxidase (GOD) was investigated. Three different molecular weighted dextrans (M(w) 17.5, 75, 188 kD) were used with different concentrations. Dramatically increased enzyme
Yonggang Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 28(8), 3831-3839 (2012-02-02)
We studied the interfacial tension between coexisting phases of aqueous solutions of dextran and polyethylene glycol. First, we characterized the phase diagram of the system and located the binodal. Second, the tie lines between the coexisting phases were determined using

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service