Skip to Content
Merck
All Photos(1)

Documents

D3759

Sigma-Aldrich

Dextran from Leuconostoc mesenteroides

average mol wt 48,000-90,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Leuconostoc mesenteroides)

form

powder

optical activity

[α]/D 199°

mol wt

average mol wt 48,000-90,000

color

white

solubility

H2O: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dextrans are used in many applications as platelet aggregants, plasma volume extenders, osmotic pressure regulators, stabilizers, organ separation media, matrix components, copolymers, microcarriers, binding agents, viscosity modifiers, antithrombotics, lubricants and physical structure components. They may be used as long hydrophilic spacer arms to improve the performance (freedom of movement) of conjugated/bound proteins. Dextrans may be derivatized for use in biosensor systems.
Dextrans of less than 60,000Da are generally considered low molecular weight dextrans. Low molecular weight dextrans are often preferred versus high molecular weight dextrans due to their viscosities, aggregation and permeation properties. High molecular weight, water-soluble, dextran polymers have been used in a wide variety of biomedical applications. Dextran (48,000-90,000) may be used to induce blood aggregation and as a viscosity plasma expander (HVPE).
Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena Pitkänen et al.
Investigative ophthalmology & visual science, 46(2), 641-646 (2005-01-27)
To determine the effects of solute molecular weight and lipophilicity on the permeability of a retinal pigment epithelium (RPE)-choroid preparation. Fresh RPE-choroid specimens from bovine eyes were placed in diffusion chambers for permeability experiments with carboxyfluorescein, fluorescein isothiocyanate (FITC)-labeled dextrans
Sahitya Katikaneni et al.
Bioanalysis, 3(21), 2419-2426 (2011-11-15)
The purpose of this study was to optimize parameters pertaining to microdialysis technique so as to make this method feasible for evaluating transdermal transport of macromolecules. Microdialysis experiments were performed in vivo using hairless rats with daniplestim as the model
Andreas M A O Pollet et al.
Micromachines, 11(1) (2020-01-08)
The vasculature plays a central role as the highway of the body, through which nutrients and oxygen as well as biochemical factors and signals are distributed by blood flow. Therefore, understanding the flow and distribution of particles inside the vasculature
Yuichi Ozaki et al.
Circulation journal : official journal of the Japanese Circulation Society, 76(4), 922-927 (2012-02-04)
Although an intracoronary frequency-domain optical coherence tomography (FD-OCT) system overcomes several limitations of the time-domain OCT (TD-OCT) system, the former requires injection of contrast media for image acquisition. The increased total amount of contrast media for FD-OCT image acquisition may
Molecular Weight Effects on the Miscibility Behavior of Dextran and Maltodextrin with Poly(vinylpyrrolidone).
Van Eerdenbrugh B, Taylor LS.
Pharmaceut. Res., doi: 10-doi: 10 (2012)

Protocols

Dextran polymer details: composed mainly of alpha-D-(1-6) linkages with varied branch lengths.

Dextran polymer details: composed mainly of alpha-D-(1-6) linkages with varied branch lengths.

Dextran polymer details: composed mainly of alpha-D-(1-6) linkages with varied branch lengths.

Dextran polymer details: composed mainly of alpha-D-(1-6) linkages with varied branch lengths.

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service