Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dextran from
Leuconostoc
spp.
Linear Formula:
(C
6
H
10
O
5
)
n
CAS Number:
9004-54-0
EC Number:
232-677-5
Products
Product Category
carbohydrates (6)
Brand
Sigma-Aldrich (6)
Color
Physical Form
Research Area
Application
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
31392
Dextran from
Leuconostoc
spp.,
M
r
450,000-650,000
Expand
View Pricing
31390
Dextran from
Leuconostoc
spp.,
M
r
~70,000
Expand
View Pricing
31389
Dextran from
Leuconostoc
spp.,
M
r
~40,000
Expand
View Pricing
31388
Dextran from
Leuconostoc
spp.,
M
r
~6,000
Expand
View Pricing
09184
Dextran from
Leuconostoc
spp.,
M
r
~100,000
Expand
View Pricing
31387
Dextran from
Leuconostoc
spp.,
M
r
15,000-25,000
Expand
View Pricing