Skip to Content
Merck
All Photos(1)

Key Documents

239M-1

Sigma-Aldrich

Collagen Type IV (CIV22) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

CIV22, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (239M-14)
vial of 0.5 mL concentrate (239M-15)
bottle of 1.0 mL predilute (239M-17)
vial of 1.0 mL concentrate (239M-16)
bottle of 7.0 mL predilute (239M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG1

control

lung

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... COL4A1(1282)

General description

Collagen Type IV is the major component of the basal lamina so antibodies to this molecule confirm its presence and reveal the morphological appearance of the structure. Normal tissue stains with this antibody in a fashion consistent with the sites of mesenchymal elements and epithelial basal laminae. Anti-Collagen IV can also be useful in the classification of soft tissue tumors: schwannomas and leiomyomas. Their well-differentiated, malignant counterparts usually immunoreact with this antibody. The vascular nature of neoplasms, hemangiopericytoma, angiosarcoma and epithelioid hemangioendothelioma can be revealed by this antibody with greater reliability than non-specific stains (e.g. silver reticulum)

Quality


IVD

IVD

IVD

RUO

Linkage

Collagen Type IV Positive Control Slides, Product No. 239S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Määttä et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 49(6), 711-726 (2001-05-25)
Laminins (Ln), together with Type IV collagen and nidogen-1, form the structural integrity of the basement membranes (BM). In this study we used immunohistochemistry to show the distribution of laminin chains alpha1, alpha3, alpha5, beta1, beta2, beta3, gamma1, gamma2, as
P De Iorio et al.
Anticancer research, 21(2B), 1395-1399 (2001-06-09)
The prognostic variability in breast cancer patients prompted the authors to investigate specific biological markers for the identification of high-risk breast cancer groups. In the present study, attention was focused on the interaction between tumor cells and the extracellular matrix
S H Barsky et al.
The American journal of surgical pathology, 7(7), 667-677 (1983-10-01)
A new method has been developed for identifying blood vessel capillaries and distinguishing them from lymphatic capillaries. Highly purified antibodies to two ubiquitous components of basement membrane--Type IV collagen and laminin--were applied to fresh-frozen and formalin-fixed tissue sections rich in
S Damiani et al.
Virchows Archiv : an international journal of pathology, 434(3), 227-234 (1999-04-06)
A retrospective study was made of 38 selected brest tumours with a poorly differentiated in situ duct component. These were classified on haematoxylin and eosin (H&E) as ductal carcinoma in situ (DCIS; 10 cases), DCIS with invasion (17 cases) and
J P McArdle et al.
International journal of cancer, 34(5), 633-638 (1984-11-15)
The basal lamina in a variety of metastatic tumours in brain was assessed with an antibody to type-IV collagen and the indirect immunoperoxidase technique. The antibody was raised in rabbits against type-IV collagen isolated from human placental tissue. Basal lamina

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service