Skip to Content
Merck
All Photos(1)

Key Documents

17348

Sigma-Aldrich

2-Hydroxyethyl methacrylate

Synonym(s):

1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate, HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OH
CAS Number:
Molecular Weight:
130.14
Beilstein:
1071583
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

vapor density

5 (vs air)

vapor pressure

0.01 mmHg ( 25 °C)

Assay

≥99%

form

liquid

contains

~0.001% hydroquinone monomethyl ether as stabilizer

impurities

<250 mg/kg free acid (as methycrylic acid) (T)

refractive index

n20/D 1.453 (lit.)
n20/D 1.453

bp

67 °C/3.5 mmHg (lit.)

density

1.071 g/mL at 20 °C
1.073 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Hydroxyethyl methacrylate has been used in a study to describe a novel methodology for grafting polymers via radical photopolymerization initiated on gold surfaces by aryl layers from diazonium salt precursors. 2-Hydroxyethyl methacrylate has also been used in a study to provide insight into the induction of apoptosis and the differential expression of antioxidant enzymes depending on the availability of glutathione.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarra Gam-Derouich et al.
Langmuir : the ACS journal of surfaces and colloids, 28(21), 8035-8045 (2012-05-03)
In this paper we describe a novel methodology for grafting polymers via radical photopolymerization initiated on gold surfaces by aryl layers from diazonium salt precursors. The parent 4-(dimethylamino)benzenediazonium salt was electroreduced on a gold surface to provide 4-(dimethylamino)phenyl (DMA) hydrogen
Stephanie Krifka et al.
Biomaterials, 33(21), 5177-5186 (2012-04-27)
Resin monomers like 2-hydroxyethyl methacrylate (HEMA) disturb cell functions including responses of the innate immune system, mineralization and differentiation, or induce cell death via apoptosis. These phenomena are associated with oxidative stress and a reduction in the concentration of the
Jie Yin et al.
Advanced materials (Deerfield Beach, Fla.), 24(40), 5441-5446 (2012-08-24)
Ordered herringbone patterns with deterministic long and short wavelengths are created using a sequential wrinkling strategy (SWS). Patterns with a prescribed zig-zag turning angle less than 90° are obtained upon sequential wrinkling of non-equi-biaxial prestrain for the first time. SWS
Anette Kocbach Bølling et al.
Toxicology letters, 216(2-3), 130-138 (2012-11-28)
Methacrylate monomers have been identified in aqueous extracts of freshly cured dental fillings. The hypothesis tested presently was that low concentrations of triethyleneglycol dimethacrylate (TEGDMA) and 2-hydroxyethyl methacrylate (HEMA) alone or in combination interfere with the LPS-induced release of cytokines
Y Yoshida et al.
Journal of dental research, 91(11), 1060-1065 (2012-09-13)
Previous research showed that the functional monomer 10-methacryloxydecyl dihydrogen phosphate (MDP) ionically bonds to hydroxyapatite (HAp) and forms a nano-layered structure at the interface with HAp-based substrates. Such hydrophobic nano-layering is considered to contribute to the long-term durability of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service