Skip to Content
Merck
All Photos(1)

Key Documents

408980

Sigma-Aldrich

2-(Diethylamino)ethyl methacrylate

contains 1500 ppm MEHQ as inhibitor, 99%

Synonym(s):

β-(Diethylamino)ethyl methacrylate, 2-(N ,N -Diethylamino)ethyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2N(C2H5)2
CAS Number:
Molecular Weight:
185.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

contains

1500 ppm MEHQ as inhibitor

refractive index

n20/D 1.444 (lit.)

bp

80 °C/10 mmHg (lit.)

density

0.922 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCOC(=O)C(C)=C

InChI

1S/C10H19NO2/c1-5-11(6-2)7-8-13-10(12)9(3)4/h3,5-8H2,1-2,4H3

InChI key

SJIXRGNQPBQWMK-UHFFFAOYSA-N

Application

2-(Diethylamino)ethyl methacrylate can be used as a precursor in the preparation of:
  • Polymeric mediator containing ferrocene moieties which are applicable in electrochemical biosensors.
  • Sepiolite-based nanocomposites by (cryo)polymerization method.
  • Thermoresponsive triblock copolymers via group transfer polymerization (GTP).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akifumi Kawamura et al.
Colloids and surfaces. B, Biointerfaces, 99, 74-81 (2011-11-15)
Bioconjugated gel particles that have complexes composed of lectin concanavalin A (ConA) and 2-glucosyloxyethyl methacrylate (GEMA) were synthesized by the surfactant-free emulsion copolymerization of N,N-diethylaminoethyl methacrylate (DEAEMA), poly(ethylene glycol) dimethacrylate (PEGDMA), GEMA, and modified-ConA with polymerizable groups. The resultant gel
Daniel Shae et al.
Nature nanotechnology, 14(3), 269-278 (2019-01-22)
Cyclic dinucleotide (CDN) agonists of stimulator of interferon genes (STING) are a promising class of immunotherapeutics that activate innate immunity to increase tumour immunogenicity. However, the efficacy of CDNs is limited by drug delivery barriers, including poor cellular targeting, rapid
Synthesis and study of copolymer of vinylferrocene, acrylamide and 2-(diethylamino)ethyl methacrylate as a polymeric mediator for electrochemical biosensors
Zuifang Liu, et al.
Reactive and Functional Polymers, 70(10), 715-725 (2010)
Sumon Ganguli et al.
Macromolecular bioscience, 10(8), 853-859 (2010-03-20)
Poly(N,N-diethylaminoethyl methacrylate)-graft-poly(ethylene glycol) (PEAMA-g-PEG) has previously been used as a novel additive to improve the heat resistance of lysozyme, which has a positive net charge and a negatively charged active site. In the present study, we show that PEAMA-g-PEG prevents
Dave J Adams et al.
Journal of controlled release : official journal of the Controlled Release Society, 128(2), 165-170 (2008-04-26)
Vesicles prepared from block copolymers have been mooted for the encapsulation of water-soluble molecules. This is because the membranes of polymer vesicles have been shown to be more stable than those in vesicles formed from lipids, with the membrane properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service