Skip to Content
Merck
All Photos(3)

Key Documents

T0501

Sigma-Aldrich

Thymol

≥98.5%

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

vapor pressure

1 mmHg ( 64 °C)

Assay

≥98.5%

form

powder or crystals

bp

232 °C (lit.)

mp

48-51 °C (lit.)

solubility

ethanol: 50 mg/mL

density

0.965 g/mL at 25 °C (lit.)

antibiotic activity spectrum

fungi

application(s)

diagnostic assay manufacturing
hematology
histology

Mode of action

DNA synthesis | interferes

storage temp.

room temp

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thymol is generally added in the Hematoxylin and Eosin staining solution to prevent mould growth. It is also suitable for use in antimicrobial screening by the disk diffusion method. It has been used as a test compound in antifungal assay to assess the susceptibility of various fungal isolates.

Disclaimer

The product is not intendedfor use as a biocide under global biocide regulations, including but notlimited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, EuropeanBiocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’sBiocidal Products Regulation, Korea’s Consumer Chemical Products and BiocideSafety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Duverger et al.
Journal of cellular physiology, 228(3), 654-664 (2012-08-14)
Mutations in DLX3 in humans lead to defects in craniofacial and appendicular bones, yet the in vivo activities related to Dlx3 function during normal skeletal development have not been fully elucidated. Here we used a conditional knockout approach to analyze
Sandeep Sharma et al.
Antimicrobial agents and chemotherapy, 53(1), 216-222 (2008-06-25)
Hydroxychavicol isolated from the chloroform extraction of aqueous extract of Piper betle leaves showed inhibitory activity against oral cavity pathogens. It exhibited an inhibitory effect on all of the oral cavity pathogens tested (MICs of 62.5 to 500 microg/ml) with
Poliana Guerino Marson Ascêncio et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 190543-190543 (2015-01-15)
Costus spiralis (Costaceae) is a species native to the Amazon region and is used in traditional medicine. The endophytic fungi used in this study were obtained from leaves of this plant. 13 strains were selected to obtain hydroethanolic extracts and
Hasan Mahmud et al.
Cardiovascular research, 98(1), 37-46 (2013-01-24)
Suicidal death of erythrocytes (eryptosis) is characterized by cell shrinkage and exposure of phosphatidylserine (PS) residues at the cell surface. Excessive eryptosis may lead to anaemia. We aimed to study the role of eryptosis in heart failure (HF)-associated anaemia. We
S P Lee et al.
British journal of pharmacology, 153(8), 1739-1749 (2008-03-13)
Thymol, a major component of thyme and oregano, has medical uses in oral care products as an astringent and antibiotic. Its distinctive sharp odour and pungent flavour are considered aversive properties. The molecular basis of these aversive properties is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service