Skip to Content
Merck
All Photos(1)

Key Documents

12006C

Sigma-Aldrich

Fetal Bovine Serum

Australia origin, USDA approved, Heat Inactivated, suitable for cell culture

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Premier

quality

USDA approved

composition

Hemoglobin, ≤25 mg/dL

origin

Australia origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL Endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Application

FBS sourced from Australia is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture. Australia carries the best possible rating in relation to Geographical Bovine Spongiform Encephalopathy (BSE) Risk (GBR) as designated by the Scientific Steering Committee on the Geographical Risk of Bovine Spongiform Encephalopathy. The classification for Australia is GBR Level I.

Preparation Note

Heat inactivated for 30 minutes at 56° C.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert C Tuckey et al.
The Journal of steroid biochemistry and molecular biology, 112(4-5), 213-219 (2008-11-13)
Cytochrome P450scc (CYP11A1) metabolizes vitamin D3 to 20-hydroxyvitamin D3 as the major product, with subsequent production of dihydroxy and trihydroxy derivatives. The aim of this study was to determine whether cytochrome P450scc could metabolize 1alpha-hydroxyvitamin D3 and whether products were
Li Zhou et al.
Genome biology, 9(7), R119-R119 (2008-07-31)
CD8+ NKT-like cells are naturally occurring but rare T cells that express both T cell and natural killer cell markers. These cells may play key roles in establishing tolerance to self-antigens; however, their mechanism of action and molecular profiles are
Ruth M Risueño et al.
PloS one, 3(3), e1747-e1747 (2008-03-06)
How the T cell antigen receptor (TCR) discriminates between molecularly related peptide/Major Histocompatibility Complex (pMHC) ligands and converts this information into different possible signaling outcomes is still not understood. One current model proposes that strong pMHC ligands, but not weak
Silvia Goldoni et al.
The American journal of pathology, 173(3), 844-855 (2008-08-09)
Decorin, a member of the small leucine-rich proteoglycan gene family, down-regulates members of the ErbB receptor tyrosine kinase family and attenuates their signaling, leading to growth inhibition. We investigated the effects of decorin on the growth of ErbB2-overexpressing mammary carcinoma
Thi A Tran et al.
Journal of neuroinflammation, 5, 14-14 (2008-05-14)
Recent animal and human studies implicate chronic activation of microglia in the progressive loss of CNS neurons. The inflammatory mechanisms that have neurotoxic effects and contribute to neurodegeneration need to be elucidated and specifically targeted without interfering with the neuroprotective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service