Merck
  • The IL-10 and IFN-gamma pathways are essential to the potent immunosuppressive activity of cultured CD8+ NKT-like cells.
The IL-10 and IFN-gamma pathways are essential to the potent immunosuppressive activity of cultured CD8+ NKT-like cells.
Genome biology (2008-07-31)
Li Zhou, Hongjie Wang, Xing Zhong, Yulan Jin, Qing-Sheng Mi, Ashok Sharma, Richard A McIndoe, Nikhil Garge, Robert Podolsky, Jin-Xiong She
ABSTRACT
CD8+ NKT-like cells are naturally occurring but rare T cells that express both T cell and natural killer cell markers. These cells may play key roles in establishing tolerance to self-antigens; however, their mechanism of action and molecular profiles are poorly characterized due to their low frequencies. We developed an efficient in vitro protocol to produce CD8+ T cells that express natural killer cell markers (CD8+ NKT-like cells) and extensively characterized their functional and molecular phenotypes using a variety of techniques. Large numbers of CD8+ NKT-like cells were obtained through culture of naïve CD8+ T cells using anti-CD3/anti-CD28-coated beads and high dose IL-2. These cells possess potent activity in suppressing the proliferation of naïve responder T cells. Gene expression profiling suggests that the cultured CD8+ NKT-like cells and the naïve CD8+ T cells differ by more than 2-fold for about 3,000 genes, among which 314 are upregulated by more than 5-fold and 113 are upregulated by more than 10-fold in the CD8+ NKT-like cells. A large proportion of the highly upregulated genes are soluble factors or surface markers that have previously been implicated in immune suppression or are likely to possess immunosuppressive properties. Many of these genes are regulated by two key cytokines, IL-10 and IFN-gamma. The immunosuppressive activities of cells cultured from IL-10-/- and IFN-gamma-/- mice are reduced by about 70% and about 50%, respectively, compared to wild-type mice. Immunosuppressive CD8+ NKT-like cells can be efficiently produced and their immunosuppressive activity is related to many surface and soluble molecules regulated by IL-10 and IFN-gamma.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture