Skip to Content
Merck
All Photos(1)

Key Documents

12306C

Sigma-Aldrich

Fetal Bovine Serum

non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture

Synonym(s):

HI FBS, HI FCS, HI Fetal Bovine Serum, HI Fetal Calf Sera, Heat Inactivated Fetal Calf Sera, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
bovine fetus
origin:
non-USA origin
sterility:
sterile-filtered

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Premier

quality

from USDA approved countries

composition

Hemoglobin, ≤25 mg/dL

origin

non-USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS sourced from USDA approved non-USA countries is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.
Fetal Bovine Serum has been used to block the human induced pluripotent stem cells (hiPSCs) derived hepatocyte-like cells (HLCs) cells in phosphate buffered saline during immunofluorescence staining.

Biochem/physiol Actions

Fetal Bovine Serum (FBS) is used as a supplement to cell culture media. It provides many non-defined growth promoting and survival enhancing factors to cells in culture.

Suitability

USDA Approved Sourcing refers to FBS produced from blood collected in countries outside of the United States that have been approved by the USDA (United States Department of Agriculture) for export of beef products into the United States. Eligible countries for export into the U.S. include Australia, Canada, Chile, Costa Rica, Honduras, Iceland, Japan, Mexico, New Zealand, Nicaragua, Uruguay. Foreign serum is tested to ensure the absence of exotic viruses before release by the USDA.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Generation of functional hepatocyte-like cells from human pluripotent stem cells in a scalable suspension culture
Vosough M, et al.
Stem Cells and Development, 22(20), 2693-2705 (2013)
Differentiation and transplantation of human induced pluripotent stem cell-derived hepatocyte-like cells
Asgari S, et al.
Stem Cell Reviews and Reports, 9(4), 493-504 (2013)
Xiao-Min Liu et al.
Nature chemical biology, 15(9), 865-871 (2019-08-07)
RNA modification in the form of N6-methyladenosine (m6A) regulates nearly all the post-transcriptional processes. The asymmetric m6A deposition suggests that regional methylation may have distinct functional consequences. However, current RNA biology tools do not distinguish the contribution of individual m6A
Shao Thing Teoh et al.
Cancer letters, 492, 21-30 (2020-08-10)
Dysregulated metabolism is a hallmark of cancer that supports tumor growth and metastasis. One understudied aspect of cancer metabolism is altered nucleotide sugar biosynthesis, which drives aberrant cell surface glycosylation known to support various aspects of cancer cell behavior including
Michael P Collins et al.
The Journal of biological chemistry, 295(28), 9433-9444 (2020-05-16)
The vacuolar H+-ATPase (V-ATPase) is an ATP-dependent proton pump that is essential for cellular homeostasis. V-ATPase activity is controlled by the regulated assembly of the enzyme from its component V1 and V0 domains. We previously reported that amino acid starvation

Articles

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

See All

Related Content

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service