Skip to Content
Merck
All Photos(1)

Documents

11039

Sigma-Aldrich

L-Arginine monohydrochloride

≥99.5% (AT), suitable for ligand binding assays, BioUltra

Synonym(s):

(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Arginine monohydrochloride, BioUltra, ≥99.5% (AT)

product line

BioUltra

Assay

≥99.5% (AT)

form

powder or crystals

optical activity

[α]20/D +22.0±0.5°, c = 5% in 5 M HCl

technique(s)

ligand binding assay: suitable

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.05%

loss

≤0.05% loss on drying, 20 °C (HV)

color

colorless

pH

5.5-7.0 (25 °C, 1 M in H2O)

mp

220-230 °C

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.42 g/cm3 at 20 °C

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.2
λ: 280 nm Amax: ≤0.1

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Arginine monohydrochloride has been used to study the reversal of the inhibitory effect of L-NG0-nitroarginine on non-cholinergic relaxations. It has also been used as a dietary supplement to study its effect on endothelium-dependent vasodilation.

Biochem/physiol Actions

L-Arginine is considered as a semi essential amino acid. In most mammals including humans, L-Arginine synthesis occurs through intestinal-renal axis, from glutamine, glutamate and proline. Abnormal levels of L-Arginine is associated with the development of kidney and cardiovascular disease. L-arginine is found to decrease fat mass and visceral adiposity.
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-Arginine and its metabolites in kidney and cardiovascular disease.
Popolo A
Amino Acids, 46(10), 2271-2286 (2014)
L-arginine for the treatment of centrally obese subjects: a pilot study.
Hurt RT
Journal of Dietary Supplements, 11(1), 40-52 (2014)
L-NG monomethyl arginine and L-NG-nitro arginine inhibit
non-adrenergic, non-cholinergic relaxation of the mouse
anococcygeus muscle
A. Gibson
British Journal of Pharmacology, 99, 602-606 (1990)
Antiatherogenic Effects of L-Arginine in the Hypercholesterolemic Rabbit
John P. Cooke
The Journal of Clinical Investigation, 90, 1168- 1172 (1992)
C B Christiansen et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(7), 537-543 (2015-01-21)
TLQP-21 is a VGF-derived neuropeptide proposed to be involved in regulation of metabolism. More specifically it has been suggested that TLQP-21 has the ability to enhance glucose stimulated insulin secretion, making it a candidate for treatment of patients with type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service