1.01542
L-Arginine
for biochemistry
Synonym(s):
L-Arginine, Arg
About This Item
Recommended Products
Quality Level
form
solid
potency
>5110 mg/kg LD50, oral (Rat)
pH
11.4 (20 °C, 100 g/L in H2O)
mp
238 °C (decomposition)
bulk density
640 kg/m3
storage temp.
2-30°C
SMILES string
[N+H3]C(CCCN=C(N)N)C(=O)[O-]
InChI
1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)
InChI key
ODKSFYDXXFIFQN-UHFFFAOYSA-N
Analysis Note
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine-grain powder or colorless crystals
Appearance of solution (0.5 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +25.0 to +26.5
UV-Absorption (260 nm; 0.5 mol/l; 1 cm; water): ≤ 0.35
UV-Absorption (280 nm; 0.5 mol/l; 1 cm; water): ≤ 0.15
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.5 %
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service