Skip to Content
Merck
All Photos(1)

Key Documents

255327

Sigma-Aldrich

Tripropyl phosphate

99%

Synonym(s):

Tri-n-propyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2O)3P(O)
CAS Number:
Molecular Weight:
224.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

120-122 °C/10 mmHg (lit.)

density

1.012 g/mL at 25 °C (lit.)

functional group

phosphate

SMILES string

CCCOP(=O)(OCCC)OCCC

InChI

1S/C9H21O4P/c1-4-7-11-14(10,12-8-5-2)13-9-6-3/h4-9H2,1-3H3

InChI key

RXPQRKFMDQNODS-UHFFFAOYSA-N

General description

Tripropyl phosphate is an organophosphorus flame retardant and plasticizer[1].

Application

Tripropyl phosphate has been used as external standard in sampling of organophosphate triesters in indoor air by using solid phase extraction cartridges[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Bacaloni et al.
Environmental science & technology, 42(6), 1898-1903 (2008-04-16)
The concentration levels, distribution, and seasonal fluctuations of 12 organophosphorus flame retardants and plasticizers (OPs), of which some are reported to be toxic to aquatic organisms, were investigated in lakes from June 2006 to June 2007. Three volcanic lakes located
Thorvald Staaf et al.
Journal of environmental monitoring : JEM, 7(4), 344-348 (2005-03-31)
In this paper we demonstrate the use of SPE cartridges for sampling of organophosphate triesters in indoor air by adsorptive enrichment. The method has been optimised for the sampling and analysis of organophosphate triesters using a 25 mg aminopropyl silica
Shaogang Chu et al.
Chemosphere, 180, 169-177 (2017-04-14)
Organophosphate (OP) and organophosphate ester (OPE) adducts of albumin are valuable biomarkers for retrospective verification of exposure. In the present study, our goal was to determine whether OPE flame retardants (OPE FRs) and OPE plasticizers can covalently bind to human
Xiaobo Zheng et al.
Chemosphere, 184, 185-191 (2017-06-09)
Indoor dust has been widely used to monitor flame retardants (FRs) in indoor environment, but most studies only focused on floor dust. In the present study, FRs were examined in indoor dust from different locations. Dust from air conditioner (AC)
Hongli Tan et al.
Environmental science & technology, 52(19), 11017-11026 (2018-09-11)
The present study investigated the occurrence of 20 organophosphate esters (OPEs) in house dust from 51 South China homes and the risks of human exposure to OPEs via two pathways: dust ingestion and hand-to-mouth contact. In addition to several traditional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service