Skip to Content
Merck
All Photos(4)

Key Documents

05470

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, crystallized, ≥98.0% (GE)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

bovine

Assay

≥98.0% (GE)

form

lyophilized powder

mol wt

~66 kDa

packaging

glass bottle of

technique(s)

cell culture | mammalian: suitable
electron microscopy: suitable

pH

4.8-7.5

solubility

H2O: slightly soluble

anion traces

chloride (Cl-): ≤2000 ppm
sulfate (SO42-): ≤1000 ppm

cation traces

Ca: ≤500 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤100 ppm
Cu: ≤20 ppm
Fe: ≤20 ppm
K: ≤50 ppm
Mg: ≤500 ppm
Mn: ≤5 ppm
Na: ≤7000 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Zn: ≤10 ppm

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Application

Albumin from bovine serum can be used for Electron and Immunoelectron microscopy.[1] It can also be used for isolating and culturing neonatal rat cardiac ventricular myocytes.[2]
Bovine serum albumin has been used to quantify proteins in raw enzyme extracts.[3][4] It has also been used as a blocking agent for mitochondrial imaging.[5]

Biochem/physiol Actions

Bovine serum albumin also refers as BSA or Fraction V is a protein isolated from cows. BSA can block vacant binding sites in enzyme-linked immunosorbent assay (ELISA) in both poly-l-lysine (PLL)-treated and non-treated microwells.[6] It can also trigger insulin dependent diabetes mellitus.[7]
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Other Notes

Review[8]
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro digestibility of specific dsRNA by enzymes of digestive tract of shrimp Litopenaeus vannamei
Alvarez-Sanchez AR, et al.
Revista MVZ Cordoba, 22(2), 5867-5880 (2017)
Glucagon-like peptide-1 inhibits vascular smooth muscle cell dedifferentiation through mitochondrial dynamics regulation
Torres G, et al.
Biochemical Pharmacology, 104(1), 52-61 (2016)
Effect of dietary chitin on digestive enzyme activity, growth and survival of Macrobrachium tenellum juvenile prawns
Santos-Romero RB, et al.
Latin American Journal of Aquatic Research, 45(1), 130-138 (2017)
T. Peters, Jr.
Adv. Prot. Chem., 37, 161-161 (1985)
J D Marsh et al.
Circulation, 98(3), 256-261 (1998-08-11)
The role of androgens in producing cardiac hypertrophy by direct action on cardiac myocytes is uncertain. Accordingly, we tested the hypothesis that cardiac myocytes in adult men and women express an androgen receptor gene and that myocytes respond to androgens

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service