Merck
Androgen receptors mediate hypertrophy in cardiac myocytes.
Circulation (1998-08-11)
J D Marsh, M H Lehmann, R H Ritchie, J K Gwathmey, G E Green, R J Schiebinger
ABSTRACT
The role of androgens in producing cardiac hypertrophy by direct action on cardiac myocytes is uncertain. Accordingly, we tested the hypothesis that cardiac myocytes in adult men and women express an androgen receptor gene and that myocytes respond to androgens by a hypertrophic response. We used reverse transcription-polymerase chain reaction methods to demonstrate androgen receptor transcripts in multiple tissues and [3H]phenylalanine incorporation and atrial natriuretic peptide secretion as markers of hypertrophy in cultured rat myocytes. Messenger RNA encoding androgen receptors was detected in myocytes of male and female adult rats, neonatal rat myocytes, rat heart, dog heart, and infant and adult human heart. Both testosterone and dihydrotestosterone produced a robust receptor-specific hypertrophic response in myocytes, determined by indices of protein synthesis and atrial natriuretic peptide secretion. Androgen receptors are present in cardiac myocytes from multiple species, including normal men and women, in a context that permits androgens to modulate the cardiac phenotype and produce hypertrophy by direct, receptor-specific mechanisms. There are clinical implications for therapeutic or illicit use of androgens in humans.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Pricing and availability is not currently available.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Pricing and availability is not currently available.