Skip to Content
Merck
All Photos(3)

Key Documents

S9625

Sigma-Aldrich

Sodium chloride

ReagentPlus®, ≥99%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

reagent

Agency

suitable for GB 5009.253-2016

vapor pressure

1.33 hPa ( 865 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder or crystals

pH

7

mp

801 °C (lit.)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium chloride can be used as:      
  • A template to synthesize 2D silicon nanosheets using tetraethyl orthosilicate (TEOS) as the silica source. The large surface area of NaCl promotes the formation of large-area SiO2 nanosheets via a self-assembly process.      
  • A template to prepare tin dioxide (SnO2) hollow nanocubes, which are applicable as anode materials for lithium-ion batteries.
  • A catalyst support in the large-scale synthesis of carbon nanofibers using metal oxalates as catalysts by a chemical vapor deposition method.

Features and Benefits

NaCl is:      
  • Highly water-soluble and cost-effective.      
  • Easily recycled via a recrystallization process.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn P Trogden et al.
Current biology : CB, 29(14), 2339-2350 (2019-07-16)
The microtubule (MT) network is an essential regulator of insulin secretion from pancreatic β cells, which is central to blood-sugar homeostasis. We find that when glucose metabolism induces insulin secretion, it also increases formation of Golgi-derived microtubules (GDMTs), notably with
Anabel Fernández-Iglesias et al.
Journal of cellular and molecular medicine, 23(2), 877-886 (2018-11-13)
Liver cells isolated from pre-clinical models are essential tools for studying liver (patho)physiology, and also for screening new therapeutic options. We aimed at developing a new antibody-free isolation method able to obtain the four main hepatic cell types (hepatocytes, liver
Christopher S Morrow et al.
Cell stem cell, 26(4), 558-568 (2020-02-29)
Maintaining a healthy proteome throughout life is critical for proper somatic stem cell function, but the complexities of the stem cell response to increases in damaged or aggregated proteins remain unclear. Here we demonstrate that adult neural stem cells (NSCs)

Questions

1–4 of 4 Questions  
  1. How many 500mg pills come in this bottle?

    1 answer
    1. This product is in powder or crystalline for and not offered in pill format. Unless otherwise stated, this product is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. How much calcium this product contain?

    1 answer
    1. This product is not tested for the presence of calcium. The minimum purity specification is 99.0%. Any calcium present would be in trace amounts. If calcium determination is critical for the intended application, please see the link below for a product that is tested for this element:
      https://www.sigmaaldrich.com/product/mm/106404

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service