Skip to Content
Merck
All Photos(4)

Key Documents

900505

Sigma-Aldrich

Sodium carbonate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

≥99.5%

form

powder

quality

free-flowing

pH

12 (25 °C, 106 g/L)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

mp

851 °C (lit.)

application(s)

PFAS testing

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Palomero-Gallagher et al.
The Journal of comparative neurology, 508(6), 906-926 (2008-04-12)
Anterior cingulate cortex (ACC) is involved in emotion, mood, and autonomic regulation. Although a subgenual part of ACC (sACC) may be vulnerable in depression and area 25 is cytologically unique, there are no assessments that contrast this region with pregenual
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known
I Irwansyah et al.
Advanced materials (Deerfield Beach, Fla.), 27(4), 648-654 (2014-12-03)
Antimicrobial hydrogels are prepared based on the co-assembly of commercial Fmoc-phenylalanine and Fmoc-leucine, which act as the hydrogelator and antimicrobial building block, respectively. This co-assembled antimicrobial hydrogel is demonstrated to exhibit selective bactericidal activity for gram-positive bacteria while being biocompatible
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service