Skip to Content
Merck
All Photos(7)

Key Documents

793639

Sigma-Aldrich

Calcium chloride

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥96%
grade:
ACS reagent
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for SM 5210

vapor pressure

0.01 mmHg ( 20 °C)

product line

Redi-Dri

Assay

≥96%

form

powder

quality

free-flowing

impurities

≤0.006 meq/g Titr. base

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium chloride can be used as a catalyst to synthesize:
  • β-Aminoketones via three-component Mannich reaction of ketones, aldehydes, and amines.
  • γ-Nitro dicarbonyl esters via Michael addition reaction.
  • Pyrroles via solvent-free Paal-Knorr reaction.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niloy Chandra Sarker et al.
Journal of environmental sciences (China), 82, 179-191 (2019-05-28)
Manure is an impending source of carbon (C), sulfur (S) and water (H2O). Consequently, microbial populations utilize these constituents to produce methane (CH4), carbon dioxide (CO2), greenhouse gases (GHGs), and hydrogen sulfide (H2S). Application of nanoparticles (NPs) to stored manure
Bin Liu et al.
Materials science & engineering. C, Materials for biological applications, 112, 110905-110905 (2020-05-16)
Three-dimensional (3D) bioprinting is an extremely convenient biofabrication technique for creating biomimetic tissue-engineered bone constructs and has promising applications in regenerative medicine. However, existing bioinks have shown low mechanical strength, poor osteoinductive ability, and lacking a suitable microenvironment for laden
Qing Chang et al.
Scandinavian journal of immunology, 92(2), e12877-e12877 (2020-03-08)
Osteoclasts are responsible for bone destruction in rheumatoid arthritis (RA), and adipose-derived mesenchymal stromal cells (ADSCs) can inhibit experimental collagen-induced arthritis model. This study aims to determine whether ADSCs also suppresses osteoclastogenesis and bone erosion in collagen-induced arthritis (CIA). Osteoclasts
Andrea F Jones et al.
Cell reports, 31(10), 107729-107729 (2020-06-11)
The medial prefrontal cortex (mPFC) modulates a range of behaviors, including responses to noxious stimuli. While various pain modalities alter mPFC function, our understanding of changes to specific cell types underlying pain-induced mPFC dysfunction remains incomplete. Proper activity of cortical
Yongqing Yang et al.
The Plant cell, 31(6), 1367-1384 (2019-04-10)
Saline-alkali soil is a major environmental constraint impairing plant growth and crop productivity. In this study, we identified a Ca2+ sensor/kinase/plasma membrane (PM) H+-ATPase module as a central component conferring alkali tolerance in Arabidopsis (Arabidopsis thaliana). We report that the

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service