Skip to Content
Merck
All Photos(1)

Documents

449709

Sigma-Aldrich

Calcium chloride

AnhydroBeads, −10 mesh, ≥99.9% trace metals basis

Synonym(s):

Calcium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.01 mmHg ( 20 °C)

product line

AnhydroBeads

Assay

≥99.9% trace metals basis

impurities

≤1000.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium Chloride (CaCl2) is a water soluble ionic crystal with a high enthalpy change of solution. It is majorly derived from limestone and is a by-product of the Solvay process. It is an anhydrous salt that has a hygroscopic nature and can be used as a desiccant.

Application

CaCl2 can be used for a variety of applications such as:
  • to decrease the freezing and to act as a de-icing agent
  • for the fabrication of an absorbent that can be used in solar absorption and dehumidification
  • can be mixed in concrete to provide a measure of the moisture

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium chloride
Kemp R and Keegan SE
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Selection of effective and efficient snow removal and ice control technologies for cold-region bridges
Zhang J, et al.
Journal of Civil, Environmental, and Architectural Engineering, 3(1), 1-14 (2009)
Activated carbon, silica-gel and calcium chloride composite adsorbents for energy efficient solar adsorption cooling and dehumidification systems
Tso CY and Chao CYH
International Journal of Refrigeration, 35(6), 1626-1638 (2012)
The system CaO-Al2O3-CaCl2-H2O at 23 ?2 ?C and the mechanisms of chloride binding in concrete
Brown P and Bothe Jr J
Cement and Concrete Research, 34(9), 1549-1553 (2004)
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service