Skip to Content
Merck
All Photos(1)

Key Documents

1.02378

Supelco

Calcium chloride

anhydrous granules Reag. Ph Eur

Synonym(s):

Calcium chloride, Calcium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-140-8
NACRES:
NB.21
Assay:
≥98.0%
form:
solid
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

Assay

≥98.0%

form

solid

potency

2630 mg/kg LD50, skin (Rat)

impurities

≤0.2% Free alkali (as Ca(OH)2)

loss

≤5.0% loss on drying, 200°C

pH

8-10 (20 °C, 100 g/L in H2O)

mp

772 °C

density

2.15 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ca+2].[Cl-].[Cl-]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Application


  • Salt pollution reduces turbidity, dissolved organic matter, and cyanobacteria in experimental vernal pool communities: Discusses environmental applications of calcium chloride, focusing on its use in managing ecosystems affected by salt pollution, demonstrating its role in environmental remediation and management. (Vigil and Schuler, 2024).

Analysis Note

Assay (calculated on dried substance): ≥ 98.0 %
Free alkali (as Ca(OH)₂): ≤ 0.2 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 25.0 %
Loss on drying (200 °C): ≤ 5.0 %
Conforms Reag. Ph.Eur.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service