Skip to Content
Merck
All Photos(2)

Key Documents

538124

Sigma-Aldrich

Propionaldehyde

reagent grade, 97%

Synonym(s):

Propanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CHO
CAS Number:
Molecular Weight:
58.08
Beilstein:
506010
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
46-50 °C (lit.)
vapor pressure:
18.77 psi ( 55 °C)
4.89 psi ( 20 °C)

grade

reagent grade

vapor density

2 (vs air)

vapor pressure

18.77 psi ( 55 °C)
4.89 psi ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

404 °F

expl. lim.

17 %, 26 °F
2.6 %, 31 °F

dilution

(for analytical testing)

impurities

≤2.5% (water)

refractive index

n20/D 1.362 (lit.)

bp

46-50 °C (lit.)

mp

−81 °C (lit.)

solubility

organic solvents: soluble

density

0.805 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)CC

InChI

1S/C3H6O/c1-2-3-4/h3H,2H2,1H3

InChI key

NBBJYMSMWIIQGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionaldehyde, also known as propanal, is commonly utilized as an intermediate in organic reactions. It can be synthesized by the oxidation of propane in gas-phase. Photodissociation behavior of propionaldehyde has been investigated by the Paneth mirror method. The degradation mechanism of propionaldehyde on Rh(111) surface has been analyzed using temperature programmed desorption (TPD) and high resolution electron energy loss spectroscopy (HREELS). 2-Çhloropropionaldehyde has been synthesized from propionaldehyde, via electrochemical chlorination. The presence of two stable rotamers in propionaldehyde has been reported based on microwave spectral data.

Application


  • Unraveling the electrocatalytic reduction mechanism of enols on copper in aqueous media.: Research on the electrocatalytic reduction mechanisms of enols, including implications for propionaldehyde, provides valuable information for chemical synthesis and industrial applications in catalysis (Cui et al., 2022).


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of 1-propanol and propionaldehyde on rhodium (111).
Brown NF and Barteau MA.
Langmuir, 8(3), 862-869 (1992)
Mechanism of the Photolysis of Propionaldehyde.
Garrison WM and Burton M.
J. Chem. Phys. , 10(12), 730-739 (1942)
Chlorination of propionaldehyde by electrolysis of aqueous chloride solutions.
Maksimov KHA, et al.
Russian Journal of Applied Chemistry, 68(4), 524-526 (1995)
Microwave spectrum of propionaldehyde.
Butcher SS and Wilson Jr EB.
J. Chem. Phys. , 40(6), 1671-1678 (1964)
Eagleson M.
Concise Encyclopedia Chemistry, 898-898 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service