Skip to Content
Merck
All Photos(1)

Documents

W292303

Sigma-Aldrich

Propionaldehyde

≥97%, FG

Synonym(s):

Propanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CHO
CAS Number:
Molecular Weight:
58.08
FEMA Number:
2923
Beilstein:
506010
EC Number:
Council of Europe no.:
90
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.002
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor density

2 (vs air)

vapor pressure

18.77 psi ( 55 °C)
4.89 psi ( 20 °C)

Assay

≥97%

form

liquid

autoignition temp.

404 °F

expl. lim.

17 %, 26 °F
2.6 %, 31 °F

refractive index

n20/D 1.362 (lit.)

bp

46-50 °C (lit.)

mp

−81 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

chocolate; cocoa; nutty; whiskey; wine-like

storage temp.

2-8°C

SMILES string

[H]C(=O)CC

InChI

1S/C3H6O/c1-2-3-4/h3H,2H2,1H3

InChI key

NBBJYMSMWIIQGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Apple, banana, blue cheese, cauliflower, cooked pork, grape brandy, potato chips, sake, tea, whiskey and wine.
Propionaldehyde is a key volatile constituent of whole Granny Smith apples, onions and ginger.

Biochem/physiol Actions

Taste at 5-10 ppm

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile Phoducts of Apples III. Identification of Aldehydes and Ketones.
Huelin FE
Australian Journal of Biological Sciences, 5(3), 328-334 (1952)
Lian Hua Luo et al.
Applied microbiology and biotechnology, 89(3), 697-703 (2010-10-05)
Although the de novo biosynthetic mechanism of 3-hydroxypropionic acid (3-HP) in glycerol-fermenting microorganisms is still unclear, the propanediol utilization protein (PduP) of Lactobacillus species has been suggested to be a key enzyme in this regard. To verify this hypothesis, a
Lian Hua Luo et al.
Bioresource technology, 103(1), 1-6 (2011-10-25)
The pduP gene encodes a propionaldehyde dehydrogenase (PduP) was investigated for the role in 3-hydroxypropionic acid (3-HP) glycerol metabolism in Klebsiella pneumoniae. The enzyme assay showed that cell extracts from a pduP mutant strain lacked measurable dehydrogenase activity. Additionally, the
Onion flavor and odor, the volatile flavor components of onions.
Carson JF & Wong FF
Journal of Agricultural and Food Chemistry, 9(2), 140-143 (1961)
Cynthia A Morgan et al.
Chemico-biological interactions, 234, 29-37 (2014-12-03)
The human aldehyde dehydrogenase (ALDH) superfamily consists of at least 19 enzymes that metabolize endogenous and exogenous aldehydes. Currently, there are no commercially available inhibitors that target ALDH1A1 but have little to no effect on the structurally and functionally similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service