Skip to Content
Merck
All Photos(2)

Documents

110132

Sigma-Aldrich

Valeraldehyde

97%

Synonym(s):

Pentanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CHO
CAS Number:
Molecular Weight:
86.13
Beilstein:
1616304
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021107
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

autoignition temp.

428 °F

refractive index

n20/D 1.394 (lit.)

bp

102-103 °C (lit.)

mp

−92 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)CCCC

InChI

1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3

InChI key

HGBOYTHUEUWSSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valeraldehyde also known as pentanal, serves as a precursor or intermediate to build more complex organic molecules.

Application

Valeraldehyde has been used to study its time-weighted average sampling using solid-phase microextraction (SPME) device.
Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds.
Valeraldehyde is used in flavoring compounds and as a rubber accelerator.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Hyun Kim et al.
Journal of separation science, 35(21), 2914-2921 (2012-10-16)
In this study, the sorptive loss patterns for volatile organic compounds were evaluated by gaseous standards containing 13 compounds (benzene, toluene, styrene, p-xylene, methyl ethyl ketone, methyl isobutyl ketone, isobutyl alcohol, butyl acetate, acetaldehyde, propionaldehyde, butyraldehyde, isovaleraldehyde, and valeraldehyde). The
Aldol condensation of cyclopentanone with valeraldehyde over metal oxides
Maki-Arvela et al.
Catalysis Letters, 149, 1383-1395 (2019)
François Fenaille et al.
Rapid communications in mass spectrometry : RCM, 18(1), 67-76 (2003-12-23)
Upon hexanal-modification in the presence of NaCNBH(3), the oxidized B chain of insulin becomes mono- and further dialkylated on both the N-terminal and Lys(29) residues. A pseudo-MS(3) study was performed with a triple-quadrupole mass spectrometer on the different modified lysine-containing
Ulrich Neuenschwander et al.
Ultrasonics sonochemistry, 19(5), 1011-1014 (2012-03-06)
The application of high-amplitude ultrasound to liquids triggers cavitation. By the collapse of the thereby appearing vacuum cavities, high temperatures can be reached in a transient manner. The high temperatures in these hot-spots can lead to homolytic scission of chemical
Trine K Dalsgaard et al.
Journal of agricultural and food chemistry, 54(17), 6367-6373 (2006-08-17)
Aldehydes formed as a result of lipid oxidation form fluorophores after binding to proteins. The structure of the fluorophores formed by reaction between saturated aldehydes and lysine has not yet been identified. The reaction products formed in the reaction between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service