Skip to Content
Merck
All Photos(2)

Key Documents

537438

Sigma-Aldrich

Propyl acetate

≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740764
EC Number:
MDL number:
UNSPSC Code:
12352108
eCl@ss:
39022103
PubChem Substance ID:
NACRES:
NA.21
bp:
102 °C (lit.)
vapor pressure:
25 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

842 °F

expl. lim.

1.7 %, 37 °F
8 %

impurities

≤0.01% Acetic acid (free acid)
≤0.1% Water

evapn. residue

≤0.01%

color

APHA: ≤15

refractive index

n20/D 1.384 (lit.)

bp

102 °C (lit.)

mp

−95 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

CCCOC(C)=O

InChI

1S/C5H10O2/c1-3-4-7-5(2)6/h3-4H2,1-2H3

InChI key

YKYONYBAUNKHLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyl acetate (n-propyl acetate) is the propyl ester of acetic acid. The absolute rate constant for the reaction of n-propyl acetate with OH radicals has been reported to be (k × 10-9lmole-1s-1) = 2.6 ± 0.5lmole-1s-1.[1] Liquid-liquid equilibrium of the ternary system composed of water, acetic acid and propyl acetate has been investigated at various temperatures (298.15, 313.15 and 363.15K). Experimental values were found to be in close agreement with that of UNIQUAC (UNIversal QUAsiChemical) model.[2] It has been identified as one of the volatile ester produced from Penicillium commune.[3]

Application

Propyl acetate may be employed as a solvent for the evaluation of vapor pressure of the polymeric solution of polystyrene.[4] It may be used in the preparation of geranyl acetate.[5]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.2 °F - closed cup

Flash Point(C)

11.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geranyl acetate synthesis by lipase-catalyzed transesterification in supercritical carbon dioxide.
Geranyl acetate synthesis by lipase-catalyzed transesterification in supercritical carbon dioxide.
Chulalaksananukul W, et al.
Enzyme and Microbial Technology, 15(8), 691-698 (1993)
Rate constants for the reaction of OH radicals with n-propyl acetate, sec-butyl acetate, tetrahydrofuran and peroxyacetyl nitrate.
Winer AM, et al.
Chemical Physics, 51(2), 221-226 (1977)
Liquid-liquid equilibria for the ternary system water+ acetic acid+ propyl acetate.
Xiao X, et al.
Journal of Chemical and Engineering Data, 51(2), 582-583 (2006)
A Sunesson et al.
Applied and environmental microbiology, 61(8), 2911-2918 (1995-08-01)
Five fungal species, Aspergillus versicolor, Penicillium commune, Cladosporium cladosporioides, Paecilomyces variotii, and Phialophora fastigiata, were cultivated on two media, malt extract agar and dichloran glycerol agar. Culture flasks provided with inlet and outlet tubes were used and purified, and humidified
A L Gustavsson et al.
Bioorganic & medicinal chemistry, 5(12), 2173-2183 (1998-02-12)
The enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate, which are conformationally constrained analogues of (Z)-5-decenyl acetate (1), a sex pheromone component of the turnip moth, Agrotis segetum, have been synthesized and tested using the electrophysiological single-sensillum technique. The analogues mimic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service