Skip to Content
Merck
All Photos(3)

Key Documents

34861

Sigma-Aldrich

2-Butanone

suitable for HPLC, ≥99.7%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39021202
PubChem Substance ID:
Assay:
≥99.7%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
80 °C (lit.)
vapor pressure:
71 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.49 (vs air)

Quality Level

vapor pressure

71 mmHg ( 20 °C)

Assay

≥99.7%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

technique(s)

HPLC: suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.1% water (Karl Fischer)

evapn. residue

≤0.001%

transmittance

335 nm, ≥50%
340 nm, ≥80%
350 nm, ≥98%

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 335 nm Amax: ≤0.30
λ: 340 nm Amax: ≤0.10
λ: 350 nm Amax: ≤0.01

application(s)

food and beverages

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Physical properties of the solutions of 1-n-butyl-3-methylimidazolium tetrafluoroborate (ionic liquid) in various organic solvents (acetonitrile, dichloromethane, 2-butanone and N,N-dimethylformamide) has been studied. Viscosity and density of these mixtures were evaluated at 298.15K.[1]

Application

2-Butanone may be employed as a solvent for the methylation agent [11C]methyl trifluoromethanesulfonate ([11C]CH3OTf), during the completely automated radiosynthesis of 2-(4-N-[11C]methylaminophenyl)-6-hydroxybenzothiazole ([11C]PIB).[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A volumetric and viscosity study for the mixtures of 1-n-butyl-3-methylimidazolium tetrafluoroborate ionic liquid with acetonitrile, dichloromethane, 2-butanone and N, N-dimethylformamide.
Wang J, et al.
Green Chemistry, 5(5), 618-622 (2003)
C Philippe et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(9), 1212-1217 (2011-05-10)
[(11)C]PIB is still the standard PET compound for Alzheimer imaging targeting beta-amyloid plaques. We aimed to establish a fully-automated procedure for the synthesis and purification of [(11)C]PIB with a high degree of reliability and improved specific activity as well as
L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene
Consalvo Petti et al.
Oncotarget, 6(1), 221-233 (2014-12-05)
Constitutively active receptor tyrosine kinases (RTKs) are known oncogenic drivers and provide valuable therapeutic targets in many cancer types. However, clinical efficacy of RTK inhibitors is limited by intrinsic and acquired resistance. To identify genes conferring resistance to inhibition of
L Schafranek et al.
Leukemia, 29(1), 76-85 (2014-05-13)
Kinase inhibitors block proliferative signals in BCR-ABL1+ leukemic cells, but their capacity to induce apoptosis is poorly understood. Initial studies suggested that very brief exposure to kinase inhibitors was sufficient to induce apoptosis in chronic myeloid leukemia (CML) cells. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service