Skip to Content
Merck
All Photos(1)

Documents

W217417

Sigma-Aldrich

Butyl acetate

natural, ≥98%, FG

Synonym(s):

n-Butyl acetate, Butyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2174
Beilstein:
1741921
EC Number:
Council of Europe no.:
194c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.004
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

790 °F

expl. lim.

7.6 %

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

banana; green; sweet

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Emission factors and source profiles of volatile organic compounds from the automobile manufacturing industry.: The study investigates the emission factors of various volatile organic compounds, including butyl acetate, from the automobile manufacturing sector, providing data crucial for environmental monitoring and regulation (You et al., 2024).

Legal Information

For R&D Use and/or for Food Manufacturing only

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Margulis-Goshen et al.
International journal of pharmaceutics, 393(1-2), 230-237 (2010-04-21)
A new composition of a fully water-dilutable microemulsion system stabilized by natural surfactants is presented as a template for preparation of celecoxib nanoparticles. Nanoparticles are obtained as a dry powder upon rapid conversion of microemulsion droplets with dissolved celecoxib into
Hala Fam et al.
Bioprocess and biosystems engineering, 35(8), 1367-1374 (2012-03-29)
The mass transfer of phenol and butyl acetate to/from water was studied in two-phase partitioning bioreactors using immiscible organic solvents and solid polymer beads as the partitioning phases in a 5-L stirred tank bioreactor. Virtually instantaneous mass transfer was observed
Subhash Bhatia et al.
Journal of hazardous materials, 164(2-3), 1110-1117 (2008-11-04)
The low concentration and high flow rate of air-borne butyl acetate (BA) could be effectively removed using combined adsorption-catalytic oxidation system. Ag-Y (Si/Al=80) dual-function adsorbent was investigated for the adsorption step of 1000 ppm of butyl acetate at gas hourly
Cheng Teng Wong et al.
Journal of hazardous materials, 157(2-3), 480-489 (2008-02-26)
The performance of silver-loaded zeolite (HY and HZSM-5) catalysts in the oxidation of butyl acetate as a model volatile organic compound (VOC) was studied. The objective was to find a catalyst with superior activity, selectivity towards deep oxidation product and
Yung-Chieh Tsao et al.
The Science of the total environment, 409(17), 3158-3165 (2011-05-31)
This study evaluated the efficacy of simultaneously employing three open-path Fourier transform infrared (OP-FTIR) spectrometers with 3-day consecutive monitoring, using an odor episode as an example. The corresponding monitoring paths were allocated among the possible emission sources of a semiconductor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service