Skip to Content
Merck
All Photos(2)

Key Documents

110264

Sigma-Aldrich

2-Butanone

ReagentPlus®, ≥99%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
80 °C (lit.)
vapor pressure:
71 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

dilution

(for general lab use)

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanone, also known as methyl ethyl ketone (MEK), is a commonly used organic solvent. Due to its low boiling point, low viscosity, ability to accommodate high solid concentrations, and wide diluent tolerance, it has wide applications in the polymer industry and as a solvent in adhesives and paints.[1]

Application


  • Lipid and Volatile Profiles of Finnish Oat Batches of Pure Cultivars: Investigates the effects of storage on volatile formation including 2-Butanone in food processing, which is significant for food scientists and researchers looking into food preservation and flavor profiling (Puganen et al., 2024).

  • Changes in Volatile Compounds and Lipid Oxidation in Various Tissues of Nile Tilapia During Ice Storage: Examines the impact of storage on the volatile compound profiles of seafood, including 2-Butanone, valuable for food scientists and technologists working with seafood preservation and quality assessment (Karbsri et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Identification of chemical interferences in aldehyde and ketone determination using dual-wavelength detection"
Analytical Chemistry, 68(19), 3354- 3358 (1996)
Ionic-liquid-supported organocatalyst: efficient and recyclable ionic-liquid-anchored proline for asymmetric aldol reaction
Miao W and Chan TH
Advanced Synthesis & Catalysis, 348(12-13), 1711-1718 (2006)
2-Butanone
Hoell D, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Chao Long et al.
Journal of hazardous materials, 203-204, 251-256 (2011-12-30)
A novel hypercrosslinked polymeric adsorbent (HY-1) with high surface area and specific bimodal pore size distribution in the regions of micropore (0.5-2.0 nm) and meso-macropore (30-70 nm) was prepared. Adsorption properties of benzene and methyl ethyl ketone (MEK) vapors onto
L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service