Skip to Content
Merck
All Photos(2)

Key Documents

32568

Supelco

Cimaterol

VETRANAL®, analytical standard

Synonym(s):

2-Amino-5-(1-hydroxy-2-isopropylaminoethyl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3O
CAS Number:
Molecular Weight:
219.28
Beilstein:
2418129
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC(C)NCC(O)c1ccc(N)c(c1)C#N

InChI

1S/C12H17N3O/c1-8(2)15-7-12(16)9-3-4-11(14)10(5-9)6-13/h3-5,8,12,15-16H,7,14H2,1-2H3

InChI key

BUXRLJCGHZZYNE-UHFFFAOYSA-N

General description

Cimaterol is a β-adrenergic agonist.[1]
Cimaterol is classified under the β-agonist class of drugs.[2]

Application

Cimaterol may be used as a reference standard for the determination of the analyte in animal tissues, feeding-stuffs, and feces by high-performance liquid chromatography and post-column derivatization.[3]
Cimaterol may be used as an analytical reference standard for the determination of the analyte in:
  • Feed samples by solid-phase extraction (SPE) and ultra-performance liquid chromatography (UPLC).[2]
  • Pig liver, kidney and muscle samples by SPE and UPLC coupled to tandem mass spectrometry (MS/MS).[4]
  • Bovine retina by SPE and gas chromatography-mass spectrometry (GC-MS).[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Schiavone et al.
Journal of animal physiology and animal nutrition, 88(3-4), 94-100 (2004-04-03)
Illegal dietary supplementation with beta(2)-agonists has been shown to increase protein deposition and decrease fat accretion in domestic animals. In poultry the metabolic and endocrine responses to beta(2)-agonists are not fully elucidated. In this trial the effects of dietary clenbuterol
M Vestergaard et al.
Journal of animal science, 72(9), 2298-2306 (1994-09-01)
The objective of this study was to determine the effect of the beta-adrenergic agonist cimaterol (CIM) on fiber characteristics, capillary supply, and metabolic enzyme activities in muscles of young Friesian bulls. Four pairs of monozygotic twins in each of three
G Van Vyncht et al.
Journal of chromatography. A, 750(1-2), 43-49 (1996-10-25)
beta-Agonists are substances used in veterinary and human medicine for the treatment of pulmonary disorders. They have found a use as growth promoters to improve meat-to-fat ratios in cattle but they are not authorized for use in the European Union.
Gas chromatography-mass spectrometry analysis of β2-agonists in bovine retina.
Hernandez-Carrasquilla M
Analytica Chimica Acta, 408(1-2), 285-290 (2000)
B L Gwartney et al.
Journal of animal science, 70(7), 2144-2150 (1992-07-01)
The response time to cimaterol (CIM), a beta-adrenergic agonist, by broiler chickens for carcass characteristics, muscle composition, muscle fiber size, catheptic enzyme activity, and tenderness was determined. Two trials were conducted in which chickens were fed a control diet (CON)

Articles

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service