Skip to Content
Merck
All Photos(4)

Key Documents

309087

Sigma-Aldrich

Potassium bromate

ACS reagent, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBrO3
CAS Number:
Molecular Weight:
167.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
crystalline powder

grade

ACS reagent

Assay

≥99.8%

form

crystalline powder

impurities

≤0.005% insolubles

pH

5.0-9.0 (25 °C, 5%)

anion traces

bromide (Br-): passes test
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.002%
Na: ≤0.01%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].[O-]Br(=O)=O

InChI

1S/BrHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

OCATYIAKPYKMPG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium bromate is a strong oxidizing agent that can be used in the bromination of deactivated aromatics and in chemical oscillating reactions.

Application

Potassium bromate can be used as:
  • An oxidant in the potentiometric determination of β-lactam drug.
  • A brominating agent for the bromination of ethylacetoacetate to dibromoethylacetoacetate.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organic preparations and procedures international, 23, 395-395 (1991)
Kinetics and Mechanism of the Oxidation of Neomenthol by Potassium Bromate in Acidic Solution.
Meena R and Sharma S.
International Journal of Pharmacological Research, 4(1), 25-33 (2014)
Yingying Xu et al.
Cell reports, 29(4), 800-809 (2019-10-24)
Cockayne syndrome (CS) is a rare genetic neurodevelopmental disorder, characterized by a deficiency in transcription-coupled subpathway of nucleotide excision DNA repair (TC-NER). Mutation of the Cockayne syndrome B (CSB) gene affects basal transcription, which is considered a major cause of
Chemiluminescence determination of potassium bromate in flour based on flow injection analysis.
Yan Z, et al.
Food Chemistry, 190, 20-24 (2016)
Gordon P McCallum et al.
Toxicology and applied pharmacology, 250(2), 147-153 (2010-10-19)
In vitro and in vivo genotoxicity tests indicate methanol (MeOH) is not mutagenic, but carcinogenic potential has been claimed in one controversial long-term rodent cancer bioassay that has not been replicated. To determine whether MeOH could indirectly damage DNA via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service