Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium bromate
Synonym(s):
Potassium bromate
Linear Formula:
KBrO
3
CAS Number:
7758-01-2
Molecular Weight:
167.00
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
309087
Potassium bromate,
ACS reagent, ≥99.8%
Expand
View Pricing
60085
Potassium bromate,
puriss. p.a., ACS reagent, ≥99.8% (dried material, RT)
Expand
View Pricing
1.04912
Potassium bromate,
for analysis (max 0,000001% Hg) EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing