Skip to Content
Merck
All Photos(5)

Key Documents

221945

Sigma-Aldrich

Potassium iodide

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor pressure

1 mmHg ( 745 °C)

Assay

≥99.0%

form

crystals

impurities

≤0.001% N compounds
≤0.005% insolubles

loss

≤0.2% loss on drying

pH

6.0-9.2 (25 °C, 5%)

mp

681 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤3 ppm
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide is a water soluble inorganic salt that can be prepared by reacting hydrogen iodide and potassium bicarbonate.[1]

Application

Potassium iodide may be used as a catalyst in the following processes: Conversion of aromatic primary amines to the corresponding nitro compounds in the presence of tert-butyl hydroperoxide as oxidant.[2] Conversion of aryl halide to aryl iodides in the presence of nickel catalysts.[3] Preparation of sulfonated oxindoles from activated alkenes and sulfonylhydrazides in water and in the presence of 8-crown-6 and tert-butyl hydroperoxide.[4]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of Aryl Iodides from Aryl Halides and Potassium Iodide by Means of Nickel Catalyst
Takagi K, et al
Chemistry Letters (Jpn), 7(2), 191-192 (1978)
Selective Oxidation of Aromatic Amines to Nitro Derivatives using Potassium Iodide-tert-Butyl Hydroperoxide Catalytic System
Reddy KR, et al
Advanced Synthesis & Catalysis, 351(1-2), 93-96 (2009)
Synthesis of sulfonated oxindoles by potassium iodide catalyzed arylsulfonylation of activated alkenes with sulfonylhydrazides in water.
<BIG>Li X, et al. </BIG>
The Journal of Organic Chemistry, 78(14), 7343-7348 (2013)
Eagleson M.
Concise Encyclopedia Chemistry, 889-889 (1994)
Alumina-supported potassium iodide as a heterogeneous catalyst for biodiesel production from soybean oil.
Xie W and Li H.
J. Mol. Catal. A: Chem., 255(1), 1-9 (2006)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service