Skip to Content
Merck
All Photos(2)

Key Documents

270644

Sigma-Aldrich

Chlorobenzene

suitable for HPLC, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
132 °C (lit.)
vapor pressure:
11.8 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

99.9%

form

liquid

autoignition temp.

1178 °F

purified by

glass distillation

expl. lim.

7.1 %

technique(s)

HPLC: suitable

impurities

≤0.01% water

evapn. residue

<0.0003%

refractive index

n20/D 1.524 (lit.)

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 288 nm Amax: 1.0
λ: 290 nm Amax: 0.40
λ: 300 nm Amax: 0.05
λ: 325 nm Amax: 0.04
λ: 360-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yayun Zhu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700614-1700614 (2018-03-30)
In the planar perovskite solar cells (PSCs), the electron transport layer (ETL) plays a critical role in electron extraction and transport. Widely utilized TiO2 ETLs suffer from the low conductivity and high surface defect density. To address these problems, for
Stefan Schade et al.
Metabolites, 9(5) (2019-05-15)
Algae are key components of aquatic food chains. Consequently, they are internationally recognised test species for the environmental safety assessment of chemicals. However, existing algal toxicity test guidelines are not yet optimized to discover molecular modes of action, which require
Guanhaojie Zheng et al.
Nature communications, 9(1), 2793-2793 (2018-07-20)
Crystal orientations in multiple orders correlate to the properties of polycrystalline materials, and it is critical to manipulate these microstructural arrangements to enhance device performance. Herein, we report a controllable approach to manipulate the facet orientation within the ABX3 hybrid
En-Chi Shen et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(1), 1901952-1901952 (2020-01-11)
Inorganic mixed-halide CsPbX3-based perovskite solar cells (PeSCs) are emerging as one of the most promising types of PeSCs on account of their thermostability compared to organic-inorganic hybrid counterparts. However, dissatisfactory device performance and high processing temperature impede their development for
Aneta Baj et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-11-30)
Vitamin E is the most active natural lipophilic antioxidant with a broad spectrum of biological activity. α-Tocopherol (α-T), the main representative of the vitamin E family, is a strong inhibitor of lipid peroxidation as a chain-breaking antioxidant. Antioxidant and antiradical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service