Skip to Content
Merck
All Photos(2)

Key Documents

256412

Sigma-Aldrich

1,2,4-Trichlorobenzene

suitable for HPLC, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3
CAS Number:
Molecular Weight:
181.45
Beilstein:
956819
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.04
grade:
HPLC grade
technique(s):
HPLC: suitable
UV/Vis spectroscopy: suitable
application(s):
food and beverages
bp:
214 °C (lit.)
vapor pressure:
1 mmHg ( 40 °C)

grade

HPLC grade

vapor density

>6 (vs air)

vapor pressure

1 mmHg ( 40 °C)

Assay

≥99%

form

solid or liquid

autoignition temp.

1060 °F

expl. lim.

6.6 %, 150 °F

technique(s)

HPLC: suitable
UV/Vis spectroscopy: suitable

impurities

<0.020% water

refractive index

n20/D 1.571 (lit.)

bp

214 °C (lit.)

mp

16 °C (lit.)

solubility

water: insoluble

density

1.454 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 308 nm Amax: 1.00
λ: 310 nm Amax: 0.50
λ: 350 nm Amax: 0.05
λ: 375-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

Clc1ccc(Cl)c(Cl)c1

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Application of electrospun polyacrylonitrile/Zn-MOF-74@GO nanocomposite as the sorbent for online micro solid-phase extraction of chlorobenzenes in water, soil, and food samples prior to liquid chromatography analysis: This article explores the use of an electrospun polyacrylonitrile/Zn-MOF-74@GO nanocomposite for the extraction of chlorobenzenes, including 1,2,4-Trichlorobenzene, from various samples. The method shows high efficiency and selectivity, making it suitable for environmental and food safety applications (Amini et al., 2021).

  • Facile synthesis and immobilization of functionalized covalent organic framework-1 for electrochromatographic separation: This research involves the synthesis and application of a functionalized covalent organic framework for the electrochromatographic separation of various compounds, including chlorobenzenes like 1,2,4-Trichlorobenzene. The method shows potential for analytical and separation sciences (Bao et al., 2021).

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of several Ni/NiAl2O4 catalysts active for the 1, 2, 4-trichlorobenzene hydrodechlorination.
Cesteros Y, et al.
Applied Catalysis. B, Environmental, 25(4), 213-227 (2000)
Sorption and cosorption of 1, 2, 4-trichlorobenzene and tannic acid by organo-clays.
Dentel SK, et al.
Water Research, 32(12), 3689-3697 (1998)
The effect of trichlorobenzene solvent geometry on the morphologies of C60 nano/microcrystals produced from solution.
Liu DD, et al.
Materials Research Bulletin, 73, 65-69 (2016)
Ernest Marco-Urrea et al.
Journal of hazardous materials, 166(2-3), 1141-1147 (2009-01-31)
The degradation of 1,2,3-, 1,3,5- and 1,2,4-trichlorobenzene (TCB) by the white-rot fungus Trametes versicolor was studied. Time course experiments showed a degradation rate of 2.27 and 2.49 nmol d(-1)mg(-1) dry weight of biomass during the first 4d of incubation in
Jie Cao et al.
The Science of the total environment, 409(11), 2336-2341 (2011-03-29)
Monodispersed carboxymethyl cellulose (CMC)-stabilized Fe-Cu bimetal nanoparticles with an average diameter of less than 20nm were successfully synthesized by a modified water-based approach. The as-resulting particles exhibit a core-shell structure and are quite uniform in size and shape. Batch experiments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service