Skip to Content
Merck
All Photos(1)

Documents

STBMFTM12

Millipore

Fluid Thioglycollate Medium

ready-to-use, bottle volume 100 mL , filling volume

Synonym(s):

Fluid Thioglycollate Media, Fluid Thioglycollate Medium (FTGM), Thioglycolate broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
eCl@ss:
32014001
NACRES:
NA.74

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of

bottle capacity

125 mL

bottle volume

100 mL , filling volume

input

sample type pharmaceutical(s)

pH

7.1 °C

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest®

shipped in

ambient

storage temp.

2-30°C

suitability

anaerobic bacteria
nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Fluid Thioglycollate Medium (FTM) is a critical component of our STERITEST® solution. Composition and growth performance of this medium comply with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. Our manufacturing approach ensures the highest level of quality for our media, improving accuracy and significantly reducing the risk of incorrect interpretation and false results. The appearance of the fluid is light-yellow, slightly opalescent and viscous liquid with a pink ring in suspension ≤1 cm. The pH of the product is 7.1 ± 0.2 and can be stored at 2-25°C. The incubation temperature varies between 30-35°C, with an incubation time of 7-14 days. The product is supplied as 12 bottles of 100 mL of medium, screw cap with septum closure that minimizes the risks of cross-contamination and optimizes the disinfection procedures.

Application

Fluid Thioglycollate Medium (FTM) is primarily used for the detection of anaerobic bacteria. However, it also enables aerobic bacterial detection. The QC organisms include Clostridium sporogenes (ATCC 19404; ATCC 11437), Staphylococcus aureus (ATCC 6538) and Pseudomonas aeruginosa (ATCC 9027). This medium is used for sterility testing by membrane filtration or direct inoculation.
Sterility testing (membrane filtration and direct inoculation)
Bioburden testing

Components

Screw cap with septum

Culture Medium

Fluid Thioglycollate

Analysis Note

Turbidity indicates growth

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service