Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2,4-Trichlorobenzene
Synonym(s):
1,2,4-Trichlorobenzene
Linear Formula:
C
6
H
3
-1,2,4-(Cl)
3
CAS Number:
120-82-1
Molecular Weight:
181.45
EC Number:
204-428-0
Products
Product Category
solvents (3)
pesticides (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
256412
1,2,4-Trichlorobenzene,
suitable for HPLC, ≥99%
Expand
View Pricing
132047
1,2,4-Trichlorobenzene,
ReagentPlus
®
, ≥99%
Expand
View Pricing
296104
1,2,4-Trichlorobenzene,
anhydrous, ≥99%
Expand
View Pricing
8.21152
1,2,4-Trichlorobenzene,
for synthesis
Expand
View Pricing
36627
1,2,4-Trichlorobenzene,
PESTANAL
®
, analytical standard
Expand
View Pricing