Skip to Content
Merck
All Photos(2)

Key Documents

244147

Sigma-Aldrich

Sodium chlorate

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO3
CAS Number:
Molecular Weight:
106.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

crystals or chunks

mp

248-261 °C (lit.)

SMILES string

[Na+].[O-]Cl(=O)=O

InChI

1S/ClHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

YZHUMGUJCQRKBT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium chlorate exhibits piezoelectric effect. On heating, it decomposes to give sodium chloride and sodium perchlorate. It can be synthesized in laboratory by reacting chlorine gas with a hot aqueous solution of sodium hydroxide or sodium carbonate.

Application

Sodium chlorate (NaClO3) has been used in the preparation of aromatic amino acid and σ32 peptide (from Drosophila melanogaster) samples for NMR analysis.
It may be used in the synthesis of chlorine dioxide and sodium perchlorate. It may also be employed as a sulfation inhibitor in the culture medium of HepG2 cells ((human hepatocellular liver carcinoma cells).

Biochem/physiol Actions

sulfation biosynthesis inhibitor

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jung Ho Lee et al.
The journal of physical chemistry. B, 117(20), 6069-6081 (2013-04-09)
NMR is an extremely powerful, yet insensitive technique. Many available nuclear polarization methods that address sensitivity are not directly applicable to low-concentration biomolecules in liquids and are often too invasive. Photochemically induced dynamic nuclear polarization (photo-CIDNP) is no exception. It
Fredrik Lanner et al.
Stem cells (Dayton, Ohio), 28(2), 191-200 (2009-11-26)
Embryonic stem (ES) cells continuously decide whether to maintain pluripotency or differentiate. While exogenous leukemia inhibitory factor and BMP4 perpetuate a pluripotent state, less is known about the factors initiating differentiation. We show that heparan sulfate (HS) proteoglycans are critical
Cristóbal Viedma et al.
Chemical communications (Cambridge, England), 47(48), 12786-12788 (2011-11-05)
A single-chirality solid phase can be obtained in boiling solutions containing a racemic mixture of left- and right-handed enantiomorphous crystals due to dissolution-crystallization cycles induced by a temperature gradient. This phenomenon provides further insights into asymmetric amplification mechanisms under presumably
J B Taylor et al.
Journal of animal science, 90(1), 381-386 (2011-08-02)
Our objective was to establish doses of orally administered NaClO(3) that reduced the presence of generic Escherichia coli in intestines of ewes and neonatal lambs managed in a shed-lambing system. Neonatal lambs (n = 32; age = 7.1 ± 1.2
Fei Wang et al.
Chemosphere, 89(8), 1009-1014 (2012-08-18)
Understanding the interaction of perfluorochemicals, persistent pollutants with known human health effects, with mineral compounds in surface water and groundwater environments is essential to determining their fate and transport. Kinetic experiments showed that adsorption equilibrium can be achieved within 48

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service