Skip to Content
Merck
All Photos(3)

Key Documents

212946

Sigma-Aldrich

Copper(I) chloride

reagent grade, 97%

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
powder
solubility:
dimethyl sulfide: partially soluble(lit.)
water: insoluble(lit.)

grade

reagent grade

Quality Level

vapor pressure

1.3 mmHg ( 546 °C)

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: copper

pH

5 (20 °C, 50 g/L)

bp

1490 °C (lit.)

mp

430 °C (lit.)

solubility

dimethyl sulfide: partially soluble(lit.)
water: insoluble(lit.)

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cu is present in +1 (cuprous) valence state in CuCl. CuCl is a semiconductor having a wide band gap. It is a promising candidate for use in silicon-based optoelectronics since its lattice resembles with silicon. It participates in various reactions with Grignard reagents.

Application

Copper(I) chloride (Cuprous chloride, CuCl) has been used to synthesize 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-ol. CuCl may be used in the stereospecific synthesis of cyclic conjugated dienes. It may be employed as a reactant to investigate the stability of copper chloride complexes in hydrothermal solutions.
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (I) Chloride.
Bertz SH, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)
Copper (I) chloride-mediated intramolecular coupling of vinyltrimethylstannane and vinyl halide functions.
Piers E and Wong T.
The Journal of Organic Chemistry, 58(14), 3609-3610 (1993)
Takayoshi Arai et al.
The Journal of organic chemistry, 73(13), 4903-4906 (2008-06-03)
A catalytic asymmetric Henry reaction has been developed with use of a sulfonyldiamine-CuCl complex as a catalyst. A series of new binaphthyl-containing sulfonyldiamine ligands (2a-h) were readily synthesized in two steps starting from commercially available chiral 1,2-diamines. The (R,R)-diamine-(R)-binaphthyl ligand
Xiaoliang Xu et al.
Organic letters, 12(5), 897-899 (2010-02-04)
Promoted by CuCl/CCl(4), a variety of sulfonyl azides and tertiary amines were successfully coupled to give sulfonyl amidine derivatives in good to excellent yields. A possible mechanism for this reaction is discussed.
Experimental study of copper (I) chloride complexing in hydrothermal solutions at 40 to 300 ?C and saturated water vapor pressure.
Xiao Z, et al.
Geochimica et Cosmochimica Acta, 62(17), 2949-2964 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service