Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper(I) chloride
Synonym(s):
Copper monochloride, Cuprous chloride, Copper(I) chloride
Linear Formula:
CuCl
CAS Number:
7758-89-6
Molecular Weight:
99.00
EC Number:
231-842-9
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
212946
Copper(I) chloride,
reagent grade, 97%
Expand
View Pricing
224332
Copper(I) chloride,
ReagentPlus
®
, purified, ≥99%
Expand
View Pricing
1.02739
Copper(I) chloride,
for analysis EMSURE
®
ACS
Expand
View Pricing
229628
Copper(I) chloride,
≥99.995% trace metals basis
Expand
View Pricing
651745
Copper(I) chloride,
AnhydroBeads
™
, ≥99.99% trace metals basis
Expand
View Pricing