Skip to Content
Merck
All Photos(4)

Key Documents

202185

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

reagent grade

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
crystals or chunks
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor pressure

40 mmHg ( 0 °C)

form

crystals or chunks

concentration

24.0-25.5% (EDTA titration)

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cobalt(II) chloride hexahydrate has a cobalt ion surrounded by a tetragonal arrangement of chloride ligands. Four water molecules occupy a square plane about cobalt while chloride ions occupy axial positions.[1] Its crystals belong to the monoclinic lattice system.[2]

Application

Cobalt(II) chloride hexahydrate may be used in the following studies:
  • For the in-situ synthesis of nickel(0) and cobalt(0) nanoclusters stabilized by poly(4-styrene sulfonic acid-co-maleic acid).[3]
  • Preparation of polyvinylidene fluoride (PVDF)-cobalt (II) chloride hexahydrate nanofiber composites, via electrospinning method.[4]
  • As catalyst for the synthesis of 2-arylbenzimidazole.[5]
  • Acetylation of alcohols with carboxylic acids to form the corresponding acylated products.[6]
  • Tosylation of aliphatic and aromatic alcohols with tosylic acid.[7]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal spectra of cobalt (II) chloride hexahydrate
Joy HW and Fogel N
The Journal of Physical Chemistry, 79(4), 345-349 (1975)
Cobalt (II) catalyzed tosylation of alcohols with p-toluenesulfonic acid.
Velusamy S, et al.
Tetrahedron Letters, 45(1), 203-205 (2004)
Cobalt (II)-catalyzed direct acetylation of alcohols with acetic acid.
Velusamy S, et al.
Tetrahedron, 61(8), 2011-2015 (2005)
Preparation of PVDF nanofiber composites for hydrogen generation from sodium borohydride.
Chinnappan A, et al.
Energy, 36(2), 755-759 (2011)
Water soluble nickel (0) and cobalt (0) nanoclusters stabilized by poly (4-styrenesulfonic acid-co-maleic acid): highly active, durable and cost effective catalysts in hydrogen generation from the hydrolysis of ammonia borane.
Metin O and Ozkar S.
International Journal of Hydrogen Energy, 36(2), 1424-1432 (2011)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hi, Is this product suitable for cell culture? Thank you. Michal

    1 answer
    1. This item has not been tested for use in cell culture. However, a cell culture product option is available. Please see the link below to review product C8661: https://www.sigmaaldrich.com/US/en/product/sigma/c8661

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service