Skip to Content
Merck
All Photos(1)

Documents

232696

Sigma-Aldrich

Cobalt(II) chloride

97%

Synonym(s):

Cobalt dichloride, Cobalt muriate, Cobaltous chloride, Dichlorocobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2
CAS Number:
Molecular Weight:
129.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

vapor pressure

40 mmHg ( 0 °C)

Assay

97%

form

powder

mp

724 °C (lit.)

SMILES string

Cl[Co]Cl

InChI

1S/2ClH.Co/h2*1H;/q;;+2/p-2

InChI key

GVPFVAHMJGGAJG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabra I Djomehri et al.
Journal of cell communication and signaling, 13(1), 129-143 (2018-12-06)
While 3D cellular models are useful to study biological processes, gel-embedded organoids have large variability. This paper describes high-yield production of large (~1 mm diameter), scaffold-free, highly-spherical organoids in a one drop-one organoid format using MCF10A cells, a non-tumorigenic breast cell
Cobalt (II) chloride catalyzed chemoselective thioacetalization of aldehydes.
De SK.
Tetrahedron Letters, 45(5), 1035-1036 (2004)
Olga Bochkova et al.
Nanomaterials (Basel, Switzerland), 10(7) (2020-07-15)
The present work introduces both synthesis of silica nanoparticles doped with CoII ions by means of differently modified microemulsion water-in-oil (w/o) and Stöber techniques and characterization of the hybrid nanoparticles (CoII@SiO2) by TEM, DLS, XRD, ICP-EOS, SAXS, UV-Vis, and UV-Vis/DR
An unusual chemoselectivity in cobalt (II) chloride catalysed cleavage of oxiranes with anilines: A highly regioselective synthesis of β-amino alcohols.
Iqbal J and Pandey A.
Tetrahedron Letters, 31(4), 575-576 (1990)
Cobalt (II) chloride catalyzed acylation of alcohols with acetic anhydride: scope and mechanism.
Iqbal J and Srivastava RR.
The Journal of Organic Chemistry, 57(7), 2001-2007 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service