Skip to Content
Merck
All Photos(1)

Key Documents

12602

Sigma-Aldrich

Potassium bicarbonate

puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

99.5-101.0% (acidimetric)

form

powder or crystals

quality

meets analytical specification of Ph. Eur., BP, USP, E501

impurities

organic volatile impurities, complies (GC)
≤0.0005% heavy metals (as Pb)
≤0.002% ammonium (NH4)

loss

≤0.1% loss on drying, 4 h (on silica gel)

pH

8.0-8.6 (20 °C, 5%)

solubility

water: soluble(lit.)

anion traces

carbonate (K2CO3): ≤2.5%
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
Na: ≤300 mg/kg
Pb: ≤5 mg/kg

suitability

complies for appearance of solution

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium bicarbonate (Potassium hydrogen carbonate) is an inexpensive, nontoxic base. It is used to regulate pH or as a reagent.

Application

Potassium bicarbonate is used:
  • In the synthesis of cyclic ammonium cation N,N-dimethyl-3,5-dimethylpiperidinium (DMDMP).
  • As a base in the preparation of glycerol carbonate and glycidol from 3-chloro-1,2-propanediol (HAL).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constance Alabert et al.
Cell reports, 30(4), 1223-1234 (2020-01-30)
Chromatin states must be maintained during cell proliferation to uphold cellular identity and genome integrity. Inheritance of histone modifications is central in this process. However, the histone modification landscape is challenged by incorporation of new unmodified histones during each cell
Relation between phosphate energy donors and incorporation of labeled amino acids into proteins.
P C ZAMECNIK et al.
The Journal of biological chemistry, 209(1), 337-354 (1954-07-01)
Lisa Ceglia et al.
The Journal of clinical endocrinology and metabolism, 94(2), 645-653 (2008-12-04)
Protein is an essential component of muscle and bone. However, the acidic byproducts of protein metabolism may have a negative impact on the musculoskeletal system, particularly in older individuals with declining renal function. We sought to determine whether adding an
Karalee Jarvis et al.
Environmental science & technology, 43(16), 6314-6319 (2009-09-15)
Storage of CO2 through mineral sequestration using olivine has been shown to produce environmentally benign carbonates. However, due to the formation of a rate-limiting reaction product layer, the rate of reaction is insufficient for large-scale applications. We report the results
K Holtenius et al.
Journal of dairy science, 91(2), 743-748 (2008-01-26)
The objective of the present study was to investigate the interactions of dietary K intake typical for forage-based diets on Mg balance in lactating dairy cows. Six lactating multiparous cows of the Swedish Red and White breed in midlactation were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service