Skip to Content
Merck
All Photos(1)

Documents

Y0001132

Valsartan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N5O3
CAS Number:
Molecular Weight:
435.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valsartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(=O)N(Cc1ccc(cc1)-c2ccccc2-c3nnn[nH]3)[C@@H](C(C)C)C(O)=O

InChI

1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1

InChI key

ACWBQPMHZXGDFX-QFIPXVFZSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valsartan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moko Zeniya et al.
Hypertension (Dallas, Tex. : 1979), 62(5), 872-878 (2013-09-11)
Na-K-Cl cotransporter isoform 1 (NKCC1) is involved in the regulation of vascular smooth muscle cell contraction. Recently, the with-no-lysine kinase (WNK)-STE20/SPS1-related proline/alanine-rich kinase (SPAK)-NKCC1 phosphorylation cascade in vascular smooth muscle cells was found to be important in the regulation of
Teun van der Bom et al.
Circulation, 127(3), 322-330 (2012-12-19)
The role of angiotensin II receptor blockers in patients with a systemic right ventricle has not been elucidated. We conducted a multicenter, double-blind, parallel, randomized controlled trial of angiotensin II receptor blocker valsartan 160 mg twice daily compared with placebo
Arjan J Kwakernaak et al.
Atherosclerosis, 226(2), 459-465 (2012-12-25)
LDL-receptor deficiency may provide a mechanism which contributes to atherogenic lipoprotein abnormalities in experimental nephrosis and in humans with glomerular proteinuria. The proprotein convertase subtilisin-kexin type 9 (PCSK9) pathway plays a key role in lipoprotein metabolism by promoting LDL-receptor degradation.
Adriaan A Voors et al.
Expert opinion on investigational drugs, 22(8), 1041-1047 (2013-05-15)
Heart failure remains a syndrome with a very high mortality rate and a poor quality of life. For patients with heart failure and a preserved ejection fraction (HFpEF), no drugs have shown to improve mortality and morbidity, and therefore novel
Henry R Black et al.
Drugs, 69(17), 2393-2414 (2009-11-17)
Valsartan is a nonpeptide angiotensin receptor antagonist that selectively blocks the binding of angiotensin II to the angiotensin II type 1 receptor. The efficacy, tolerability and safety of valsartan have been demonstrated in large-scale studies in hypertension, heart failure (HF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service