Skip to Content
Merck
All Photos(1)

Key Documents

Y0000648

Telmisartan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

telmisartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)-c4ccccc4C(O)=O)-c5nc6ccccc6n5C

InChI

1S/C33H30N4O2/c1-4-9-30-35-31-21(2)18-24(32-34-27-12-7-8-13-28(27)36(32)3)19-29(31)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)

InChI key

RMMXLENWKUUMAY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Telmisartan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Telmisartan is a non-peptide AT1 angiotensin receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Li Li et al.
Diabetes, 62(3), 762-774 (2012-12-15)
The mechanisms of the improvement of glucose homeostasis through angiotensin receptor blockers are not fully elucidated in hypertensive patients. We investigated the effects of telmisartan on insulin signaling and glucose uptake in cultured myotubes and skeletal muscle from wild-type and
Mahshid Dehghan et al.
Circulation, 126(23), 2705-2712 (2012-12-06)
Diet quality is strongly related to cardiovascular disease (CVD) incidence, but little is known about its impact on CVD events in older people at high risk of CVD and receiving effective drugs for secondary prevention. This study assessed the association
Maurizio Destro et al.
Expert opinion on pharmacotherapy, 12(17), 2719-2735 (2011-11-15)
The modulation of the renin angiotensin aldosterone system (RAAS) is an important pathway in managing high blood pressure, and its overexpression plays a key role in target end-organ damage. Telmisartan is an angiotensin II receptor blocker (ARB) with unique pharmacologic
Dominik Wincewicz et al.
Pharmacological reports : PR, 66(3), 436-441 (2014-06-07)
The potential effect of chronic treatment with telmisartan, an angiotensin type 1 receptor blocker (ARB) and partial agonist of peroxisome proliferator--activated receptor γ (PPARγ), on stress-related disorders is a matter of considerable interest. The existing data suggest that angiotensin II
Anna Olverling et al.
Clinical science (London, England : 1979), 125(9), 433-438 (2013-05-30)
Diabetic patients are often treated with an ACEi (angiotensin-converting enzyme inhibitor) or angiotensin receptor antagonist against hypertension or albuminuria. These drugs also have a positive impact on glucose tolerance, but the mechanism for this remains elusive. Hypothesizing a positive non-additive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service