Skip to Content
Merck
All Photos(1)

Key Documents

1708762

USP

Valsartan

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N5O3
CAS Number:
Molecular Weight:
435.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valsartan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(=O)N(Cc1ccc(cc1)-c2ccccc2-c3nnn[nH]3)[C@@H](C(C)C)C(O)=O

InChI

1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1

InChI key

ACWBQPMHZXGDFX-QFIPXVFZSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valsartan USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Valsartan Tablets
  • Amlodipine and Valsartan Tablets
  • Valsartan and Hydrochlorothiazide Tablets
  • Amlodipine, Valsartan, and Hydrochlorothiazide Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenfeng Zheng et al.
Journal of clinical hypertension (Greenwich, Conn.), 12(6), 414-421 (2010-07-02)
This was a systematic assessment of the efficacy and safety of telmisartan and valsartan for the management of blood pressure (BP) in patients with essential hypertension. The authors reviewed randomized controlled trials (RCTs) and quasi-RCTs comparing telmisartan and valsartan for
Adriaan A Voors et al.
Expert opinion on investigational drugs, 22(8), 1041-1047 (2013-05-15)
Heart failure remains a syndrome with a very high mortality rate and a poor quality of life. For patients with heart failure and a preserved ejection fraction (HFpEF), no drugs have shown to improve mortality and morbidity, and therefore novel
Genyang Cheng et al.
Nephrology (Carlton, Vic.), 20(2), 77-84 (2014-11-02)
The current standard treatment for IgA nephropathy relies on steroid and/or immunosuppressive therapy and angiotensin converting enzyme inhibitors (ACEI) or angiotensin receptor blocker (ARB). This study examines the benefits and safety of combining valsartan with clopidogrel and leflunomide as a
Roberto Fogari et al.
Expert opinion on drug safety, 10(2), 295-303 (2010-12-15)
Angiotensin receptor blockers (ARBs) as a class are generally considered safe and better tolerated than other antihypertensive drugs. The purpose of this report is to review the main data on the safety and tolerability of the second generation ARB valsartan
Erdal Tan Ishizuka et al.
Investigative ophthalmology & visual science, 56(1), 74-80 (2014-12-17)
Extranodal marginal zone B-cell lymphoma (EMZL) is the most common subtype of conjunctival lymphoma, though its molecular mechanisms of pathogenesis are largely unknown. We attempted to explore the association of the renin-angiotensin system (RAS) and (pro)renin receptor ([P]RR) in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service