Skip to Content
Merck
All Photos(1)

Key Documents

T0010000

Talampicillin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O6S · HCl
CAS Number:
Molecular Weight:
517.98
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

talampicillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S1[C@H]2N([C@H](C1(C)C)C(=O)OC4OC(=O)c5c4cccc5)C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3.[Cl-].[H+]

InChI

1S/C24H23N3O6S.ClH/c1-24(2)17(22(31)33-23-14-11-7-6-10-13(14)21(30)32-23)27-19(29)16(20(27)34-24)26-18(28)15(25)12-8-4-3-5-9-12;/h3-11,15-17,20,23H,25H2,1-2H3,(H,26,28);1H/t15-,16-,17+,20-,23?;/m1./s1

InChI key

PAZSYTCTHYSIAO-WVFSJLEKSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Talampicillin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A double blind comparison of talampicillin and ampicillin in the treatment of exacerbations of chronic bronchitis in general practice.
H N Williams et al.
The British journal of clinical practice, 35(4), 147-152 (1981-04-01)
[Influence of antibiotics on anti-inflammatory activity of anti-inflammatory drugs in simultaneous administration].
M Yoshizawa
Nichidai koku kagaku = Nihon University journal of oral science, 13(1), 34-43 (1987-03-01)
K Fujita et al.
The Japanese journal of antibiotics, 38(8), 2149-2154 (1985-08-01)
Forty-three cases were studied to determine the most optimal timing to discontinue an oral antibacterial agent for preventing urinary tract infection after transurethral prostatectomy. On the 5th postoperative day intravenous antibiotic treatment was replaced by oral medication of 1.5 g
[Clinical evaluation of Yamacillin 250mg in dental infections].
Shikai tenbo = Dental outlook, 58(2), 383-391 (1981-08-01)
T Hashimoto et al.
The Japanese journal of antibiotics, 34(6), 942-945 (1981-06-01)
The oral administration of 300 mg of clindamycin was undertaken on 23 patients, of 500 mg of cefadroxil on 11 patients and of 250 mg of talampicillin on 12 patients, and then tooth extraction was performed under local anesthesia. Blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service