Skip to Content
Merck
All Photos(3)

Documents

P9641

Sigma-Aldrich

Nonaethylene glycol monododecyl ether

nonionic surfactant

Synonym(s):

C12E9, Dodecyl nonaethylene glycol ether, Dodecylnonaglycol, Polidocanol, Polyoxyethylene (9) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)9(CH2)11CH3
CAS Number:
Molecular Weight:
582.81
Beilstein:
1895308
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

mol wt

micellar average mol wt 64,000

aggregation number

110

CMC

0.08

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C30H62O10/c1-2-3-4-5-6-7-8-9-10-11-13-32-15-17-34-19-21-36-23-25-38-27-29-40-30-28-39-26-24-37-22-20-35-18-16-33-14-12-31/h31H,2-30H2,1H3

InChI key

ONJQDTZCDSESIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonaethylene glycol monododecyl ether [C12H25O(CH2CH2O)9H] (AEO9) is a non-ionic surfactant.

Application

Nonaethylene glycol monododecyl ether has been used in a study to assess a procedure for analyzing total lipid and triacylglycerol using bovine liver tissue. It has also been used in a study to investigate how particle size of aggregates can be measured by the concentration dependence of the amplitude of the autocorrelation function.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Starke et al.
Journal of animal science, 88(8), 2741-2750 (2010-03-30)
A procedure is described for analyzing total lipid (TL) and triacylglycerol (TAG) in 2 sequential steps using small amounts (<100 mg) of bovine liver tissue. The TL was measured gravimetrically and TAG was measured enzymatically in the TL extract, using
Jeffrey T Jensen et al.
Contraception, 94(5), 527-533 (2016-10-25)
Our objective was to conduct a pilot study to determine if transcervical administration of polidocanol foam (PF) with or without doxycycline or benzalkonium chloride (BZK) would prevent pregnancy in baboons. In study phase 1, adult cycling baboons underwent a hysterosalpingogram
Weican Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 25(4), 2363-2368 (2009-01-24)
In the pH 2.6 and 5.2 systems, soybean peroxidase (SBP) (isoelectric point, pI 3.9) has positive and negative charge, respectively. In order to acquire detailed knowledge on the role played by electrostatics in the denaturation of proteins, a comparison of
Michael C Rudolph et al.
Journal of mammary gland biology and neoplasia, 21(3-4), 131-138 (2016-11-01)
Accurate assessment of the long chain polyunsaturated fatty acid (LC-PUFA) content of human milk (HM) provides a powerful means to evaluate the FA nutrient status of breastfed infants. The conventional standard for FA composition analysis of HM is liquid extraction
Kamendra P Sharma et al.
The journal of physical chemistry. B, 115(29), 9059-9069 (2011-06-24)
We investigate the dispersion of a pH responsive polymer, polyethyleneimine, PEI, in a hexagonal (H(1)) mesophase of a nonionic surfactant, C(12)E(9), and water, at pH ranging from basic (pH = 12.8) to acidic (pH = 1). While the C(12)E(9)/H(2)O phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service